missing translation for 'onlineSavingsMsg'
Learn More

Iodine, Certified, 0.1000N ±0.0005N (0.05M), LabChem™

Catalog No. p-5379526
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC156601 500 mL Amber Glass
LC156602 1 L Amber Glass
LC156604 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC156601 Supplier LabChem Supplier No. LC156601
Only null left

Chemical Identifiers

CAS 7553-56-2 , 7681-11-0 , 7681-11-0 , 7647-01-0
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Color Brown-Red
CAS Max % 1.27, 96.17, 2.54, 0.02
Physical Form Liquid
Quantity 500 mL
Linear Formula I2
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
Solubility Information Soluble in water
Formula Weight 253.809
Grade Certified
Concentration or Composition (by Analyte or Components) 0.1000N ±0.0005N (0.05M)
Identification Passes Test
Packaging Amber Glass
Chemical Name or Material Iodine
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Avoid breathing mist, vapors, spray.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If on skin: Wash skin with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature