missing translation for 'onlineSavingsMsg'
Learn More

Iodine, 0.0315 Normal, Reagents

Catalog No. 01334915
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334915 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334915 Supplier Labchem Inc Supplier No. CI2017004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Iodine, 0.0315 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7681-11-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Diiodine solution
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Dark Brown
Physical Form Liquid
Quantity 4 L
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.01
Chemical Name or Material Iodine, 0.0315 Normal