missing translation for 'onlineSavingsMsg'
Learn More

Iodine, 0.0262 Normal, Reagents

Catalog No. 01334944
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334944 1 L
01334943 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334944 Supplier Labchem Inc Supplier No. CI0914001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Iodine, 0.0262 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7681-11-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Diiodine solution
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Brown
Physical Form Liquid
Quantity 1 L
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.01
Chemical Name or Material Iodine, 0.0262 Normal