missing translation for 'onlineSavingsMsg'
Learn More

Iodide Standard For Ion Chromatography, SPEX CertiPrep™

Catalog No. ASI92Y
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ASI92Y 125 mL Triple-leached LDPE bottle
ASI92X 500 mL Triple-leached LDPE bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ASI92Y Supplier SPEX CertiPrep Group LLC Supplier No. ASI92Y

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
I-1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7681-82-5 , 7732-18-5
Molecular Formula INa
Molecular Weight (g/mol) 149.894
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 99.9, 0.1
Analyte or Component Names Sodium iodide,distilled water,conductivity or of similar purity
Technique Ion Chromatography
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
Type Certified Reference Material
Element Sodium Iodide
Solvent or Matrix Water
Quantity 125 mL
Concentration or Composition (by Analyte or Components) 1000mg/L
Chemical Name or Material Iodide Standard, For Ion Chromatography
Show More Show Less
DOTInformation Hazardous
Recommended Storage Ambient

Product intended for laboratory use only