missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zero Blunt™ PCR Cloning Kit, without competent cells

Catalog No. K275040
Encompass
Change view
Click to view available options
Quantity:
20 Reactions
40 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K275040 40 Reactions
K275020 20 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K275040 Supplier Invitrogen™ Supplier No. K275040
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Zero Blunt™ PCR cloning kits contains: linearized pCR™-Blunt vector, ExpressLink™ T4 DNA ligase , 5X ExpressLink™ T4 DNA ligase buffer, control template, dNTPs, sterile water, and M13 forward and reverse primers. Competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.

Offers an easy method for high-efficiency (>80%) cloning of blunt-end PCR products amplified with proofreading, thermostable DNA polymerases

The Zero Blunt™ PCR Cloning Kit uses the multipurpose cloning vector pCR™-Blunt and ExpressLink™ T4 DNA Ligase to generate a ligation product in a five-minute, room-temperature ligation step.
The pCR-Blunt vector provides:

  • EcoR I sites flanking the PCR product insertion site for excision of inserts
  • T7 promoter/primer site for in vitro RNA transcription and sequencing
  • M13 forward and reverse primer sites for sequencing or PCR screening
How Zero Blunt PCR Cloning Works
The Zero Blunt PCR Cloning Kit is designed to clone blunt PCR fragments (or any blunt DNA fragment) with a low background of non-recombinants. The pCR-Blunt vector contains the lethal E. coliccdB gene fused to the C-terminus of LacZα (Bernard et al., 1994). Ligation of a blunt PCR fragment disrupts expression of the lacZα-ccdB gene fusion permitting growth of only positive recombinants upon transformation. Cells that contain non-recombinant vector are killed when the transformation mixture is plated.
Kit Configurations
The Zero Blunt PCR Cloning Kit is offered in a variety of configurations: with One Shot™ TOPO10 Chemically Competent E. coli (K2700-20 and K2700-40) and without competent cells (K2750-20 and K2750-40) in 20- and 40- reaction kit sizes.

  • Fast and convenient: 5-minute, room-temperature ligation
  • Efficient: ccdB gene for positive selection results in >80% clones with correct insert
  • Flexible: choice of kanamycin or Zeocin™ resistance for flexible antibiotic selection

Cloning, PCR Cloning

Order Info

Shipping Condition: Dry Ice

Specifications

Bacterial or Yeast Strain Not Included
Cloning Method Blunt PCR
Content And Storage Zero Blunt™ PCR cloning kits contain linearized pCR™-Blunt vector, ExpressLink™ T4 DNA ligase , 5X ExpressLink™ T4 DNA ligase buffer, control template, dNTPs, sterile water, and M13 forward and reverse primers.

Store all components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Format Kit
Product Line TOPO, Zero Blunt
Product Type PCR Cloning Kit
Promoter T7
Quantity 40 Reactions
Vector Blunt DNA Cloning Vectors
Which of the following products will usually give the best cloning efficiency in terms of positive clones and number of colonies: Zero Blunt PCR, Zero Blunt PCR-TOPO, or Zero Blunt PCR-TOPO for Sequencing?

Typically 200-300 colonies are obtained for the TOPO kits with 100% inserts (20/20). For the Zero Blunt kit that uses standard ligase, typically 600-1000 colonies are obtained with 95-100% inserts.

Is it possible to generate nested deletions using the TOPO Cloning Kit for Sequencing and the ZeroBlunt TOPO PCR Cloning Kits?

Yes. Both vectors (pCR4 and pCR-Blunt II) are designed to allow the creation of nested deletions for sequencing large inserts using the same sequencing primer. In addition, a comprehensive protocol is provided in the TOPO manual for these kits.

For Research Use Only. Not for use in diagnostic procedures.