missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zero Blunt™ PCR Cloning Kit, without competent cells

Catalog No. K275020
Encompass
Change view
Click to view available options
Quantity:
20 Reactions
40 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K275020 20 Reactions
K275040 40 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K275020 Supplier Invitrogen™ Supplier No. K275020

Includes

Zero Blunt™ PCR cloning kits contains: linearized pCR™-Blunt vector, ExpressLink™ T4 DNA ligase , 5X ExpressLink™ T4 DNA ligase buffer, control template, dNTPs, sterile water, and M13 forward and reverse primers. Competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.

Offers an easy method for high-efficiency (>80%) cloning of blunt-end PCR products amplified with proofreading, thermostable DNA polymerases

The Zero Blunt™ PCR Cloning Kit uses the multipurpose cloning vector pCR™-Blunt and ExpressLink™ T4 DNA Ligase to generate a ligation product in a five-minute, room-temperature ligation step.
The pCR-Blunt vector provides:

  • EcoR I sites flanking the PCR product insertion site for excision of inserts
  • T7 promoter/primer site for in vitro RNA transcription and sequencing
  • M13 forward and reverse primer sites for sequencing or PCR screening
How Zero Blunt PCR Cloning Works
The Zero Blunt PCR Cloning Kit is designed to clone blunt PCR fragments (or any blunt DNA fragment) with a low background of non-recombinants. The pCR-Blunt vector contains the lethal E. coliccdB gene fused to the C-terminus of LacZα (Bernard et al., 1994). Ligation of a blunt PCR fragment disrupts expression of the lacZα-ccdB gene fusion permitting growth of only positive recombinants upon transformation. Cells that contain non-recombinant vector are killed when the transformation mixture is plated.
Kit Configurations
The Zero Blunt PCR Cloning Kit is offered in a variety of configurations: with One Shot™ TOPO10 Chemically Competent E. coli (K2700-20 and K2700-40) and without competent cells (K2750-20 and K2750-40) in 20- and 40- reaction kit sizes.

  • Fast and convenient: 5-minute, room-temperature ligation
  • Efficient: ccdB gene for positive selection results in >80% clones with correct insert
  • Flexible: choice of kanamycin or Zeocin™ resistance for flexible antibiotic selection

Cloning, PCR Cloning

Order Info

Shipping Condition: Dry Ice

Specifications

Bacterial or Yeast Strain Not Included
Cloning Method Blunt PCR
Content And Storage Zero Blunt™ PCR cloning kits contain linearized pCR™-Blunt vector, ExpressLink™ T4 DNA ligase , 5X ExpressLink™ T4 DNA ligase buffer, control template, dNTPs, sterile water, and M13 forward and reverse primers.

Store all components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Format Kit
Product Line TOPO, Zero Blunt
Product Type PCR Cloning Kit
Promoter T7
Quantity 20 Reactions
Vector Blunt DNA Cloning Vectors
What is the best molar ratio of PCR product:vector to use for TOPO TA cloning? Is there an equation to calculate the quantity to use?

We suggest starting with a molar ratio of 1:1 (insert:vector), with a range of 0.5:1 to 2:1. The quantity used in a TOPO cloning reaction is typically 5-10 ng of a 2 kb PCR product.

Equation:

length of insert (bp)/length of vector (bp) x ng of vector = ng of insert needed for 1:1 (insert:vector ratio)

What is the best ratio of insert:vector to use for cloning? Is there an equation to calculate this?

The optimal ratio is 1:1 insert to vector. Optimization can be done using a ratio of 0.5-2 molecules of insert for every molecule of the vector.

Equation:

length of insert (bp)/length of vector (bp) x ng of vector = ng of insert needed for 1:1 insert:vector ratio

I'm seeing a lot of vector-only colonies when I try to perform a negative control reaction using vector only (no insert) for a TOPO reaction. Is my TOPO vector re-ligating?

Using the vector only for transformation is not a recommended negative control. The process of TOPO-adaptation is not a 100% process, therefore, there will be “vector only” present in your mix, and colonies will be obtained.

I'm trying to clone in my phosphorylated PCR product into a TOPO vector, and I'm getting no colonies. However, when I clone the same product into a TA vector, everything works perfectly. Why is this?

Phosphorylated products can be TA cloned but not TOPO cloned. This is because the necessary phosphate group is contained within the topoisomerase-DNA intermediate complex of the vector. TOPO vectors have a 3' phosphate to which topoisomerase is covalently bound and a 5' phosphate. Non-TOPO linear vectors (TA and Blunt) have a 3' OH and a 5' phosphate. Phosphorylated products should be phosphatased (CIP) before TOPO cloning.

I'm able to get a lot of colonies, however, none contain my insert of interest. What should I do?

You may be cloning in an artifact. TA and TOPO Cloning are very efficient for small fragments (< 100 bp) present in certain PCR reactions. Gel-purify your PCR product using either a silica-based DNA purification system or electroelution. Be sure that all solutions are free of nucleases (avoid communal ethidium bromide baths, for example.)

A majority of colonies are blue or light blue, with very few white colonies. What should I do?

There could be a few possibilities for this:

- The insert does not interrupt the reading frame of the lacZ gene. If you have a small insert (< 500 bp), you may have light blue colonies. Analyze some of these blue colonies as they may contain insert.
- A polymerase that does not add 3' A-overhangs was used. If you used a proofreading enzyme, you will need to do a post-reaction treatment with Taq polymerase to add the 3' A-overhangs.
- PCR products were gel-purified before ligation. Gel purification can remove the single 3' A- overhangs. Otherwise, optimization of your PCR can be performed so that you can go directly from PCR to cloning.
- The PCR products were stored for a long period of time before ligation reaction. Use fresh PCR products. Efficiencies are reduced after as little as 1 day of storage.
- Too much of the amplification reaction was added to the ligation. The high salt content of PCR can inhibit ligation. Use no more than 2-3 µl of the PCR mixture in the ligation reaction.
- The molar ratio of vector:insert in the ligation reaction may be incorrect. Estimate the concentration of the PCR product. Set up the ligation reaction with a 1:1 or 1:3 vector:insert molar ratio.
On a typical plate there are a few white colonies which do not contain insert. These are usually larger than the other colonies and are due to a deletion of a portion of the plasmid sequence by a rare recombination event (usually from the polylinker to a site in the F1 origin). To find a colony with an insert it is best to pick clones of a variety of color and pattern for analysis. Often an insert will generate two distinct patterns according to its orientation.

I'm getting no colonies after transformation. What should I do?

No colonies may occur due to the following problems:

Bacteria were not competent. Use the pUC18 vector included with the One Shot module to check the transformation efficiency of the cells.
- Incorrect concentration of antibiotic on plates, or the plates are too old. Use 100 µg/mL of ampicillin or 50 µg/mL kanamycin. Be sure ampicillin plates are fresh (< 1 month old).
- The product was phosphorylated (TOPO cloning only). Phosphorylated products can be TA-cloned but not TOPO-cloned. This is because the necessary phosphate group is contained within the topoisomerase-DNA intermediate complex of the vector. The TOPO vector has a 3' phosphate to which topoisomerase is covalently bound and a 5' phosphate. The non- TOPO vectors (TA and Blunt) have a 3' OH and a 5' phosphate. Phosphorylated products should be phosphatased (CIP) before TOPO-cloning.

I'm getting low cloning efficiency with my TOPO cloning reactions. What should I do?

Please consider the following possible causes:
- pH > 9: Check the pH of the PCR amplification reaction and adjust with 1 M Tris-HCl, pH 8.
- Excess (or overly dilute) PCR product: Reduce (or concentrate) the amount of PCR product.
- Incomplete extension during PCR: Be sure to include a final extension step of 7 to 30 minutes during PCR. Longer PCR products will need a longer extension time.
- Cloning large inserts (>1 kb): Try one or all of the following suggestions: Increase amount of insert. Incubate the TOPO cloning reaction longer. Gel-purify the insert using either a silica-based DNA purification system (e.g., PureLink system) or electroelution. Be sure that all solutions are free of nucleases (avoid communal ethidium bromide baths, for example.)
- PCR product does not contain sufficient 3' A-overhangs even though you used Taq polymerase: Increase the final extension time to ensure all 3' ends are adenylated. Taq polymerase is less efficient at adding a nontemplate 3' A next to another A. Taq is most efficient at adding a nontemplate 3' A next to a C. You may have to redesign your primers so that they contain a 5' G instead of a 5´ T.

I'm getting very few colonies after transformation of my TOPO cloning reaction. How can I increase the number of primary colonies?

Please try the suggestions below to increase the number of colonies.
- Longer incubation of the TOPO cloning reaction at room temperature, provided that the 6X Salt solution is added to the reaction.
- Electroporation can give significant increases in colony numbers; often 10-20 fold higher. However, if doing electroporation, it is important that the TOPO reaction mix contains diluted Salt solution or, for best results, precipitated prior to transformation. For high primary transformants by electroporation it is recommended to:
- Add 100 µL double diH2O to the 6 µL TOPO reaction and incubate 10 more minutes at 37 degrees C.
- Precipitate by adding 10 µL 3 M Na-Acetate, 2 µL 20 µg/µL glycogen, 300 µL 100% ethanol. Place on dry ice or –80 degrees C for 20 min, spin at top speed in a microcentrifuge at 4 degrees C for 15 min. Wash pellet with 800 µL 80% ethanol, spin at top speed for 10 min, pour off ethanol, spin 1 min, and remove remaining ethanol without disturbing pellet. Dry pellet (air-dry or speed-vac).
- Resuspend pellet in 10 µL ddH2O and electroporate 3.3 µL of resuspended DNA according to a normal electroporation protocol. This electroporation protocol can yield up to 20 fold more colonies than chemical transformation of an equivalent TOPO-reaction. The addition of the 100 µL ddH2O followed by 10 mins incubation is not absolutely necessary, but it sufficiently dilutes the reaction and may help inactivate topoisomerase so that it is more easily electroporated.

I'm planning on cloning a 1kb fragment for sequencing and want to minimize the amount of vector sequence in my data. Which of your vectors should I use?

We would suggest using our TOPO TA cloning kit for sequencing, which contains the pCR 4 TOPO vector, or our Zero Blunt TOPO PCR cloning kit for sequencing, which contains the pCR4Blunt-TOPO vector.

I have a blunt-ended restriction product cut from another cloning vector. Can I insert this into a Zero Blunt TOPO vector?

Yes, though you will need to treat it with calf intestinal phosphatase (CIP) to get rid of the 5' phosphate group for TOPO Cloning.

Which PCR polymerases do you recommend for TA/Blunt/D-TOPO cloning and why?

TA Cloning:
- This cloning method was designed for use with pure Taq polymerases (native, recombinant, hot start); however, High Fidelity or Taq blends generally work well with TA cloning. A 10:1 or 15:1 ratio of Taq to proofreader polymerase will still generate enough 3' A overhangs for TA cloning.
- Recommended polymerases include Platinum Taq, Accuprime Taq, Platinum or Accuprime Taq High Fidelity, AmpliTaq, AmpliTaq Gold, or AmpliTaq Gold 360.

Blunt cloning:
- Use a proofreading enzyme such as Platinum SuperFi DNA Polymerase.

Directional TOPO cloning:
- Platinum SuperFi DNA Polymerase works well.

Which of the following products will usually give the best cloning efficiency in terms of positive clones and number of colonies: Zero Blunt PCR, Zero Blunt PCR-TOPO, or Zero Blunt PCR-TOPO for Sequencing?

Typically 200-300 colonies are obtained for the TOPO kits with 100% inserts (20/20). For the Zero Blunt kit that uses standard ligase, typically 600-1000 colonies are obtained with 95-100% inserts.

Is it possible to generate nested deletions using the TOPO Cloning Kit for Sequencing and the ZeroBlunt TOPO PCR Cloning Kits?

Yes. Both vectors (pCR4 and pCR-Blunt II) are designed to allow the creation of nested deletions for sequencing large inserts using the same sequencing primer. In addition, a comprehensive protocol is provided in the TOPO manual for these kits.


For Research Use Only. Not for use in diagnostic procedures.