missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Yeast tRNA
Supplied in lyophilized form
$364.00 - $487.00
Specifications
| Product Type | Yeast tRNA |
|---|---|
| Content And Storage | Store in freezer (-5 to -30°C). |
| Reagent Type | Yeast tRNA |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet or Dry Ice |
| For Use With (Application) | Carrier for Nucleic Acid Precipitation |
Description
Yeast tRNA is suitable for use as a carrier in nucleic acid purification and precipitation procedures.
Array CGH, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Microarray Analysis
Order Info
Shipping Condition: Approved for shipment at Room Temperature or on Wet or Dry Ice
Specifications
| Yeast tRNA | |
| Yeast tRNA | |
| Carrier for Nucleic Acid Precipitation |
| Store in freezer (-5 to -30°C). | |
| Approved for shipment at Room Temperature or on Wet or Dry Ice | |
| Lyophilized Powder |
For Research Use Only. Not for use in diagnostic procedures.