missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Wash Buffer (25X)

Catalog No. WB01
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WB01 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. WB01 Supplier Invitrogen™ Supplier No. WB01

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suitable for use with all ELISA assays

  • Clear, colorless concentratred liquid buffer
  • Available in two sizes
  • Detection method: Colormetric

Build Your Own Immunoassay, Cell Analysis, Immunoassays

Order Info

Shipping Condition: Wet Ice

Specifications

Quantity 100 mL
Content And Storage Buffer. Store at 2°C to 8°C.
Shipping Condition Wet Ice