missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Tris Buffer (powder format)

Catalog No. 15504020
Encompass
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15504020 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15504020 Supplier Invitrogen™ Supplier No. 15504020

Includes

One 1kg container UltraPure Tris buffering agent

No DNase, RNase, or protease activity detected

  • Used because of its buffering range (pH 7.0 to 9.0) and compatibility with many enzymes, including restriction endonucleases and DNA-modifying enzymes

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Approved for shipment at room temperature or on wet ice

Specifications

Chemical Name or Material Tris
Content And Storage Contents: One container (1 kg) of UltraPure™ Tris buffering agent
For Use With (Application) Nucleic Acid Gel Electrophoresis, Blotting
Product Line UltraPure
Product Type Tris Buffer
Quantity 1 kg
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Powder

For Research Use Only. Not for use in diagnostic procedures.