missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UltraPure™ TBE Buffer, 10X

Compatible with agarose gels
$110.00 - $604.00
Specifications
| Buffer | Running Buffer |
|---|---|
| Concentration | 10 X |
| Format | Bottle |
| Gel Compatibility | Polyacrylamide Gels, Agarose Gels |
| For Use With (Application) | Nucleic Acid Gel Electrophoresis, Blotting |
Description
UltraPure 10X TBE Buffer is a sterile-filtered solution of 1M Tris, 0.9 M boric acid, and 0.01M EDTA used to prepare 1X buffer for polyacrylamide and agarose gel electrophoresis.
- UltraPure 10X TBE Buffer is available in 1 L plastic bottle/10 L Cubitainer™ box
- Performance and quality testing: No DNase, RNase, or protease activity detected. Sustainable packaging
Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
| Running Buffer | |
| Bottle | |
| Nucleic Acid Gel Electrophoresis, Blotting | |
| TBE Buffer |
| 10 X | |
| Polyacrylamide Gels, Agarose Gels | |
| UltraPure | |
| Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.