missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UltraPure™ SSPE, 20X
For use in nucleic acid hybridizations and blot transfer applications
$144.00 - $144.00
Specifications
| Concentration | 20 X |
|---|---|
| Content And Storage | Store at room temperature. |
| For Use With (Application) | Nucleic Acid Gel Electrophoresis, Blotting |
| Product Line | UltraPure |
| Shipping Condition | Room Temperature |
Description
- UltraPure 20X SSPE is prepared buffer concentrate
- Supplied in 1 L plastic bottles/4 L/10 L stackable Cubitainer™, Box
- Sterile-filtered UltraPure 20X SSPE contains 3.0 M NaCl, 0.2M NaH2PO4, and 0.02M EDTA at pH 7.4
- Performance and Quality Testing: No DNase or RNase activity detected. Sustainable packaging
DNA & RNA Purification & Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis & Blotting, Southern Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
| 20 X | |
| Nucleic Acid Gel Electrophoresis, Blotting | |
| Room Temperature |
| Store at room temperature. | |
| UltraPure | |
| Liquid |
For Research Use Only. Not for use in diagnostic procedures.