missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Guanidine Isothiocyanate

Catalog No. 15535016
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15535016 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15535016 Supplier Invitrogen™ Supplier No. 15535016

UltraPure Guanidine Isothiocyanate is a strong protein denaturant when used at high concentrations.

UltraPure Guanidine Isothiocyanate is a strong protein denaturant when used at high concentrations. It is useful in the isolation of intact RNA from sources rich in RNase such as pancreas.

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store at room temperature.
For Use With (Application) RNA Extraction
Product Type Guanidine Isothiocyanate
Product Line UltraPure
Purity DNase-Free, RNase-Free
Quantity 500 g
Shipping Condition Room Temperature
Form Powder

For Research Use Only. Not for use in diagnostic procedures.