missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Glycerol

Catalog No. 15514011
Encompass
Change view
Click to view available options
Quantity:
500 mL
3.8 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15514011 500 mL
15514029 3.8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15514011 Supplier Invitrogen™ Supplier No. 15514011

UltraPure Glycerol is suitable for the concentration and storage of enzymes at low temperatures. A 50% (v/v) solution will not freeze at -20°C.

UltraPure Glycerol (≥99% pure) is suitable for concentrating and storing enzymes at low temperatures. It can be diluted to a 50% (v/v) solution that will not freeze at -20°C. With a density of 1.26 g/mL, it is commonly used in electrophoresis loading buffers. Additionally, UltraPure Glycerol gradients are ideal for purifying proteins, bacteriophage, and organelles.

Performance and quality testing: no DNase, RNase, or protease activity detected

Order Info

Shipping Condition: Room temperature

Specifications

Chemical Name or Material Glycerol
Content And Storage Store at room temperature.
Product Line UltraPure
Purity Molecular Biology Grade
Quantity 500 mL
Shipping Condition Room Temperature
Form Liquid
What is the storage media for the competent cells?

Preparation procedures and formulations for all of our competent cells are proprietary. All chemically competent cells are delivered in an aqueous solution that contains a mixture of salts, along with a freezing stabilizer such as glycerol or DMSO.

For Research Use Only. Not for use in diagnostic procedures.