missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ DNA Typing Grade™ 50X TAE Buffer
GREENER_CHOICE

Catalog No. 24710030
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24710030 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 24710030 Supplier Invitrogen™ Supplier No. 24710030

Most commonly used buffer for agarose DNA electrophoresis

UltraPure DNA Typing Grade™ 50X TAE Buffer is a sterile-filtered solution of 2M Tris-acetate and 50 mM EDTA. To prepare a 1X solution, mix one volume of UltraPure 50X TAE Buffer with 49 volumes of distilled water.

  • Contains 40mM Tris-acetate and 1mM EDTA with a pH of 8.3 at 23°C.
  • Performance and Quality Testing: No DNase activity detected
  • Compatible with agarose and E-Gel™ gels
  • Sustainable packaging

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Room Temperature

Specifications

Buffer Running Buffer
Concentration 50 X
Content And Storage Store at room temperature.
Gel Compatibility E-Gel™, Agarose Gels
For Use With (Application) Nucleic Acid Gel Electrophoresis, Blotting
Product Line UltraPure
Product Type TAE Buffer
Quantity 1 L
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.