missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOPO™ TA Cloning™ Kit, Dual Promoter, with One Shot™ TOP10 chemically competent E. coli cells
For fast, efficient cloning and subsequent in vitro transcription/translation
$728.00 - $2,265.00
Specifications
| Bacterial or Yeast Strain | TOP10 |
|---|---|
| Cell Type | Chemically Competent |
| Cloning Method | TOPO™-TA |
| Content And Storage | Box 1: • Topoisomerase I-activated pCR™II-TOPO™ vector • PCR buffer • Salt solution • dNTPs • Control template • M13 forward and reverse primers • Control PCR primers • Sterile water Store at -5 to -30°C. All reagents are stable for 6 months when properly stored. Box 2: • One Shot™ Chemically Competent or Electrocomp™ E. coli • S.O.C. medium • Supercoiled pUC19 control plasmid. Store at -68 to -85°C. |
| Format | Kit |
Includes: Topoisomerase I-activated pCR™II-TOPO™ vector; PCR buffer; Salt solution; dNTPs; Control template; T3, T7, M13 forward, and M13 reverse primers; Control PCR primers; Sterile water; One Shot™ Chemically Competent or Electrocomp™ E. coli; S.O.C. medium; Supercoiled pUC19 control plasmid
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
K4600J10
|
Invitrogen™
K4600J10 |
10 Reactions |
Each for $728.00
|
|
|||||
K460001
|
Invitrogen™
K460001 |
25 Reactions |
Each for $1,264.00
|
|
|||||
K460040
|
Invitrogen™
K460040 |
50 Reactions |
Each for $2,265.00
|
|
|||||
Description
By eliminating time-consuming and tedious restriction site cloning, TOPO cloning is the most reliable method, taking only 5 minutes using a 3-step protocol, and yielding up to 95% recombinants. This kit includes the pCR™II-TOPO TA vector.
- 3'-T overhangs for direct ligation of Taq-amplified PCR products
- T7 and SP6 promoters for efficient in vitro transcription
- M13 forward and reverse primer sites for sequencing
- 16 convenient restriction sites, including EcoRI, flanking your insert for subsequent excision or subcloning
- Kanamycin and ampicillin resistance for your choice of selection in E. coli
- Easy blue/white colony screening for selection of recombinants
- Includes OneShot™ TOP10 Competent cells for added convenience and highest cloning efficiencies
Applications:
Cloning, PCR Cloning
Specifications
| TOP10 | |
| TOPO™-TA | |
| Kit | |
| T7, SP6 |
| Chemically Competent | |
| Box 1: • Topoisomerase I-activated pCR™II-TOPO™ vector • PCR buffer • Salt solution • dNTPs • Control template • M13 forward and reverse primers • Control PCR primers • Sterile water Store at -5 to -30°C. All reagents are stable for 6 months when properly stored. Box 2: • One Shot™ Chemically Competent or Electrocomp™ E. coli • S.O.C. medium • Supercoiled pUC19 control plasmid. Store at -68 to -85°C. |
|
| Cloning Kit |
For Research Use Only. Not for use in diagnostic procedures.