missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Terminal Deoxynucleotidyl Transferase (TdT) Buffer

Catalog No. 16314015
Encompass
Change view
Click to view available options
Quantity:
2 x 0.8 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16314015 2 x 0.8 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Hazard Warning: Toxic; potassium cacodylate contained in reaction buffer. Also contains cobalt chloride, a highly toxic chemical. See MSDS.
Catalog No. 16314015 Supplier Invitrogen™ Supplier No. 16314015

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hazard Warning: Toxic; potassium cacodylate contained in reaction buffer. Also contains cobalt chloride, a highly toxic chemical. See MSDS.

Supplied with Terminal Deoxynucleotidyl Transferase (TdT)

  • Formulation: 500mM potassium cacodylate (pH 7.2), 10mM CoCl2 , 1mM DTT

Homopolymer tailing of vector and insert for cloning; Labeling oligonucleotides with biotin, 32P- or 35S-label (3); Apoptosis (TUNEL); Cloning; Restriction Enzyme Cloning

Order Info

Shipping Conditions: Approved for shipment on wet or dry ice

Specifications

Content And Storage Terminal Deoxynucleotidyl Transferase, Recombinant (rTdT) is supplied with vial of 5X buffer [500 mM potassium cacodylate (pH 7.2), 10 mM CoCl2 , 1 mM DTT]. Store at -20°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Compatible Buffer 5X Buffer, Reaction Buffer
Quantity 2 x 0.8 mL
Product Type TdT

For Research Use Only. Not for use in diagnostic procedures.