missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Terminal Deoxynucleotidyl Transferase, recombinant

Catalog No. 10533073
Encompass
Change view
Click to view available options
Quantity:
3 x 500 U
500 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10533073 3 x 500 U
10533065 500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Hazard Warning: Toxic; potassium cacodylate contained in reaction buffer. Also contains cobalt chloride, a highly toxic chemical. See MSDS.
Catalog No. 10533073 Supplier Invitrogen™ Supplier No. 10533073
Hazard Warning: Toxic; potassium cacodylate contained in reaction buffer. Also contains cobalt chloride, a highly toxic chemical. See MSDS.

Includes

Vial of 5X buffer [500mM potassium cacodylate (pH 7.2), 10mM CoCl2 , 1mM DTT]

Performance and quality tested

Terminal Deoxynucleotidyl Transferase, Recombinant (rTdT) is DNA polymerase that catalyzes addition of deoxynucleotides to 3fi hydroxyl terminus of DNA.

  • Source: Purified from baculovirus clone of calf thymus TdT
  • Performance and Quality Testing: Endonuclease, 3' and 5' exodeoxyribonuclease, and levels of incorporation tested
  • Unit Definition: One unit incorporates 1nmol dATP into acid-precipitable material in 1 hr. at 37°C, using d(pA)50 as primer
  • Unit Reaction Conditions: 0.2M potassium cacodylate (pH 7.2), 10mM MgO4 C4 H6 , 1mM 2-mercaptoethanol, 0.5mg/mL BSA, 100 flM d(pA)50, 1mM [3H]dATP, and enzyme in 0.15mL for 1 h at 37°C

Cloning; Restriction Enzyme Cloning; Homopolymer tailing of vector and insert for cloning; Labeling oligonucleotides with biotin, 32P- or 35S-label; Apoptosis (TUNEL)

Order Info

Shipping Conditions: Approved for shipment on wet or dry ice

Specifications

Content And Storage Terminal Deoxynucleotidyl Transferase, Recombinant (rTdT) is supplied with vial of 5X buffer [500 mM potassium cacodylate (pH 7.2), 10 mM CoCl2 , 1 mM DTT]. Store at -20°C.
Enzyme Terminal Deoxynucleotidyl Transferase
Compatible Buffer 5X Buffer, Reaction Buffer
Quantity 3 x 500 U
Product Type TdT
Is the Terminal Deoxynucleotidyl Transferase (TdT) within the Pierce Biotin 3' End DNA Labeling Kit (Cat. No. 89818) available separately?

Yes, but not the specific one from that same kit. We have the following comparable Terminal Deoxynucleotidyl Transferases (Cat. No. 10533065, 10533073 or EP0161, EP0162).

For Research Use Only. Not for use in diagnostic procedures.