missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TA Cloning™ Kit, with pCR™2.1 Vector and One Shot™ INVαF' Chemically Competent E. coli

Catalog No. p-4924885
Encompass
Change view
Click to view available options
No. of Reactions:
20 Reactions
40 Reactions
Vector:
pCR2.1
pCRII
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions Vector
K200001 20 Reactions pCRII
K200040 40 Reactions pCR2.1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K200001 Supplier Invitrogen™ Supplier No. K200001
Only null left

Quick, one-step cloning strategy for directly inserting Taq-amplified PCR product into plasmid vector

The TA Cloning Kit uses the pCR2.1 cloning vector and ExpressLink T4 DNA Ligase to generate a ligation product in a 15-minute, room-temperature ligation step. Reactions typically yield >80% recombinants containing inserts.

  • Fast and convenient: 15-minute, room-temperature ligation
  • Efficient: blue/white screening and >80% clones with correct insert
  • Flexible: choice of kanamycin or ampicillin resistance for flexible antibiotic selection
  • Hassle-free: eliminates any enzymatic modifications of PCR product
  • Streamlined: does not require use of PCR primers that contain restriction sites
  • 3'-T overhangs for direct ligation of Taq-amplified PCR products
  • T7 promoter for in vitro RNA transcription and sequencing
  • Versatile polylinker with flanking EcoR I sites for easy excision of inserts
  • M13 forward and reverse primer sites for sequencing

  • Taq polymerase has non-template-dependent activity that adds single deoxyadenosine (A) to the 3' ends of PCR products
  • Linearized vector supplied in has single 3' deoxythymidine (T) residues
  • Allows PCR inserts to ligate efficiently with vector

Cloning, PCR Cloning

Specifications

Bacterial or Yeast Strain INVαF ́
Cell Type Chemically Competent E. coli
Cloning Method TA Cloning
Content And Storage TA Cloning™ kits contain linearized pCR™2.1 vector, ExpressLink™ T4 DNA ligase, 5X ExpressLink™ T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls. The competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.

Store One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Format Kit
For Use With (Application) PCR Cloning
No. of Reactions 20 Reactions
Product Line One Shot
Product Type Cloning Kit
Promoter T7
Quantity 20 reactions
Vector pCRII
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.