missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ROX Reference Dye

Catalog No. 12223012
Encompass
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12223012 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12223012 Supplier Invitrogen™ Supplier No. 12223012
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ROX Reference Dye is designed to normalize the fluorescent reporter signal in real-time quantitative PCR or RT-PCR.

  • Normalizes non-PCR-related fluctuations in fluorescence
  • Provides a stable baseline for multiplex quantitative PCR and RT-PCR
  • Uses existing ROX spectral settings, eliminating the need to re-calibrate the thermal cycler

Order Info

Shipping Conditions: Wet or Dry Ice

Specifications

Content And Storage Store in freezer (-5° to -30°C).
Detection Method Primer-probe
PCR Method qPCR
Label or Dye ROX
Product Type ROX Reference Dye
Quantity 500 μL
Shipping Condition Approved for shipment on Wet or Dry Ice
Volume 500 μL

For Research Use Only. Not for use in diagnostic procedures.