missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ribonuclease H

Catalog No. 18021071
Encompass
Change view
Click to view available options
Quantity:
120 U
30 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18021071 120 U
18021014 30 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18021071 Supplier Invitrogen™ Supplier No. 18021071

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Ribonuclease H containing 30 units supplied at a concentration of 2U/μL

Specifically degrades RNA strand of RNA-DNA hybrid to produce 5' phosphate-terminated oligoribonucleotides and single-stranded DNA

  • Source: Purified from E. coli expressing the E. coli RNase H gene on a plasmid
  • Performance and quality testing: Ribonuclease, nonspecific endodeoxyribonuclease, 3' and 5' exodeoxyribonuclease
  • Unit definition: One unit hydrolyzes 1nmol of RNA in 3H-labeled poly(A)poly(dT) to acid-soluble material in 20 min. at 37°C
  • Unit reaction conditions: 20mM Tris-HCl (pH 7.5), 0.1M KCl, 10mM MgCl2 , 0.1mM DTT, 5% (w/v) sucrose, 0.5nmol 3H-labeled poly(A)poly(dT), and enzyme in 50μL for 20 min. at 37°C

Removal of mRNA during second-strand cDNA synthesis; Removal of poly(A) sequences from mRNA in the presence of oligo(dT); Oligodeoxyribonucleotide-directed cleavage of RNA; PCR & Real-Time PCR; Reverse Transcription

Order Info

Shipping Conditions: Approved for shipment on wet or dry ice

Specifications

Concentration 2 U/μL
Content And Storage 4 vials of Ribonuclease H with each containing 30 units supplied at a concentration of 2U/μl

Store at -20°C in a non-frost-free freezer.
Shipping Condition Approved for shipment on Wet or Dry Ice
Enzyme RNase
Quantity 120 U
Product Type Ribonuclease H
Do you offer RNAse H from the SuperScript III First Strand Synthesis System as a stand-alone item?

Yes, we do offer E. coli RNAse H as a stand-alone item: Cat. Nos. 18021014 and 18021071.

Which components of the SuperScript III First Strand Synthesis System for RT-PCR are available for purchase separately?

The following components are available as stand-alone items:

- Superscript III Reverse Transcriptase (Cat. Nos. 18080093, 18080044, 18080085)
- Oligo (dT)20 Primer (Cat. No. 18418020)
- Random hexamers (Cat. No. 48190011)
- 10 mM dNTP Mix (Cat. Nos. 18427013, 18427088)
- RNAseOUT Recombinant Ribonuclease Inhibitor (Cat. No. 10777019)
- E. coli RNAse H (Cat. Nos. 18021014, 18021071)

For Research Use Only. Not for use in diagnostic procedures.