Learn More
Invitrogen™ PureLink™ Quick Gel Extraction Kit

Description
Includes
Gel Solubilization Buffer (L3)
Wash Buffer (W1)
Elution Buffer (E5); (10 mM Tris-HCl, pH 8.5)
Quick Gel Extraction Columns
Wash Tubes
Recovery Tubes
The PureLink™ Quick Gel Extraction Kit allows you to rapidly and efficiently purify DNA fragments from TAE or TBE agarose gels of various percentages. DNA can be extracted and purified from agarose gels with different melting points in ∼30 minutes using PureLink™ silica membrane-based quick gel extraction columns. For your convenience, purification protocols are provided for centrifugation and for vacuum.
Advantages of using PureLink™ Quick Gel Extraction Kit:
- Purify DNA fragments from TAE and TBE agarose gels of various percentages and melting points
- Complete the procedure in ∼30 minutes
- Easily purify DNA fragments from 40 bp to 10 kb from gels
- Obtain high recovery of DNA fragments
- Bind and purify up to 15 μg DNA with one column
- Purify DNA fragments that are high quality and show reliable performance in PCR, restriction enzyme digestion, cloning, and labeling
Note: The PureLink™ Quick Gel Extraction Kit is not designed to purify supercoiled plasmid DNA or genomic DNA from agarose gels. Only linear DNA fragments may be purified from gels using this kit.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | PureLink Elution Tubes For better long-term performance, it is recommended to store the purification columns at 2°C to 8°C. |
| Isolation Technology | Silica Spin Column |
| Sample Type | Agarose Gels, Gel Samples |
| Elution Volume | 50 μL |
| Final Product Type | DNA |
| For Use With (Application) | PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| Label or Dye | Ethidium Bromide, SYBR Safe |
| Quantity | 50 Preps |
| Shipping Condition | Room Temperature |
| Show More |
Frequently Asked Questions (FAQs)
Here are some suggestions for your experiments:
- Many enzymes, including restriction endonucleases and ligases, are inhibited by small amounts of agarose and by perchlorate contaminants. This is not normally a problem. If it is, however, you can use the DNA without repurification by increasing the amount of enzyme used in the digest or ligation or by increasing the digestion incubation time. Also, you may use less DNA in your digest or ligation with the same total volume and the same amount of restriction enzyme.
- There may have been residual ethanol in the eluted fragment. Be sure to thoroughly centrifuge to remove the wash buffer, discard the wash buffer, and use a fresh tube to collect the eluted DNA. For applications that are very sensitive to ethanol, let the open spin column stand for 15 minutes at room temperature to let any excess ethanol evaporate.
- The washing steps may not be as efficient as they should be. Under these circumstances, there may be trace amounts of perchlorate in the eluate. To avoid this, extend the centrifugation times to 5 minutes and wash 2 times with wash buffer.
- Be sure to perform the optional wash step if you are using higher concentrations of agarose or are adding more than 250 mg to the cartridge. If applications are sensitive to EDTA, elute with water (pH 7.5-8.5), or with 10 mM Tris, pH 8.0 without EDTA.
Both regular and low-melting agaroses can be used. When the agarose concentration is above 2%, use 600 µL of solubilization buffer for each 100 mg of gel.
Yes, water may be used, but please ensure that the water is clean and the pH of the water is between 7.5 and 8.5.
Yes, both TAE and TBE agarose gels are compatible with our gel extraction kits.
Multiple bands can occur when the DNA is partially denatured on the gel due to high heat generated during electrophoresis or running the gel too fast. This can appear as multiple bands when the eluted DNA is analyzed on a gel. Denaturation can also occur in AT-rich DNA during the 50 degrees C incubation to dissolve the gel slices. If this happens, solubilize the gel at 37 degrees C for 20 to 30 minutes with repeated vortexing.
For Research Use Only. Not for use in diagnostic procedures.