missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ PCR Purification Binding Buffer B2

Catalog No. K310005
Encompass
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K310005 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K310005 Supplier Invitrogen™ Supplier No. K310005
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PureLink™ PCR Purification Binding Buffer B2 is the binding buffer used with the PureLink™ PCR Purification Kit.

PureLink™ PCR Purification Binding Buffer B2 is the binding buffer used with the PureLink™ PCR Purification Kit. It is added directly to PCR products in the first step of the PureLink™ PCR Purification Kit procedure to facilitate DNA binding to the silica membrane. The buffer is activated by the addition of 80mL of Isopropanol.

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store at room temperature.
Product Line PureLink
Product Type PCR Purification Binding Buffer B2
Quantity 120 mL
Shipping Condition Room Temperature
Can I use the PureLink PCR Purification Kit to clean up restriction digested plasmid DNA?

Yes, you can use our kit to clean up restriction digested plasmid DNA.

Can I use the PureLink PCR Purification Kit to do a gel purification?

The buffers and procedure are different for the PureLink PCR Purification Kit and PureLink Gel Purification Kit. Therefore, you can swap the column, but the right buffer and protocol must be used.

What are the minimum and maximum size ranges for DNA fragment cleanup using the PureLink PCR Clean-Up Kit?

The size range is between 100 bp and 12 kb. Our kit comes with two buffers, Binding Buffer (B2) and Binding Buffer HC (B3). The Binding Buffer HC (B3) eliminates primer-dimers and short failed PCR products that are smaller than 300 bp.

For Research Use Only. Not for use in diagnostic procedures.