missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ HiPure Plasmid Midi/Maxiprep Precipitators

Catalog No. K210020S
Encompass
Change view
Click to view available options
Quantity:
4 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K210020S 4 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K210020S Supplier Invitrogen™ Supplier No. K210020S
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Traditional DNA precipitation method of anion exchange isolated plasmid DNA involves isopropanol precipitation followed by centrifugation and drying of DNA which is time consuming and labor intensive.

Traditional DNA precipitation method of anion exchange isolated plasmid DNA involves isopropanol precipitation followed by centrifugation and drying of DNA which is time consuming and labor intensive.

  • PureLink HiPure Precipitator allows plasmid DNA concentration from bacterial cultures within 5 minutes using anion exchange resin without need for centrifugation and risk of losing DNA pellet during supernatant removal
  • Recovery is >80% and resulting plasmid DNA is ready to use in the most challenging applications such as transfection

Not high throughput compatible (manual)

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store at room temperature.
Isolation Technology Anion Exchange Resin
Sample Type Bacterial Culture
Final Product Type Plasmid DNA
For Use With (Application) Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
High-throughput Compatibility Not High-throughput Compatible (Manual)
Prep Scale 100-200 μg (Medium-Scale) Plasmid DNA
Product Line PureLink
Product Type Plasmid Midi/MaxiPrep Precipitator
Quantity 4 Preps
Shipping Condition Room Temperature
Target Plasmid DNA
Test Time 5 min.
Show More Show Less
My plasmid yield was lower than expected. What could be the cause?

There are several reasons lower yields can occur:

-Make sure the binding of the plasmid is being done at RT. Temperature affects the pH of the binding solution.

-Make sure all other solutions are also warmed to RT for optimal yield.

-Verify that the centrifugation immediately following the neutralization step was not done at 4 degrees C. If it was, the supernatant MUST be warmed to RT before loading onto the column.

-The 1:1:1 ratio of buffers (Resuspension/Lysis/Precipitation) has not been kept precisely. A surplus of solution E3 will exceed the optimal salt concentration for DNA binding, thus leading to decreased DNA yield.

-Low-copy number plasmids give lower yields.

-If all of the medium was not removed at the cell harvesting step, the pH of the subsequent step can be affected and reduce yield.

-The cell pellet was not thoroughly resuspended before lysis.

-The purified DNA pellet was overdried after isopropanol precipitation and ethanol wash making it difficult to resuspend the pellet. Air dry only.

-Pellets can be easily lost during the alcohol precipitation and wash steps. It is best to pipette off the alcohol solutions rather than pouring them off, as the pellets tend to be slippery.

-After lysis and neutralization, the lysate is not homogeneous. After addition of solutions (Resuspension/Lysis/Precipitation buffers), cells must be mixed thoroughly by inverting the tubes until a homogeneous phase is obtained. Otherwise, cells will not be lysed completely and the plasmid DNA will stick to bacterial debris. In the case of bigger bacterial cell pellets, more vigorous shaking may be required to achieve homogeneity. However, do NOT vortex, as vortexing will release chromosomal DNA.

I did not recover any DNA. Why?

The DNA pellet from precipitation with isopropanol is easily dislodged when washing with 70% ethanol. It is best to remove the isopropanol supernatant and the ethanol wash by pipetting. Be careful not to shoot the washing buffer directly onto the pellet. Instead, allow the washing buffer to run over the pellet. Regardless of which manufacturer's miniprep kit you use, washing the pellet can be challenging because it is so small.

What is the binding capacity of the PureLink HiPure Precipitator?

The binding capacity of the PureLink HiPure Precipitator is 2 mg.

Can I use the PureLink HiPure Precipitator with other anion exchange kits?

For best results, we recommend the use of the PureLink HiPure Precipitators with our PureLink HiPure Plasmid Filter Maxiprep/Midiprep Kit or PureLink HiPure Plasmid Maxiprep/Midiprep kit. However, PureLink HiPure Precipitator can be used with an equivalent anion exchange plasmid purification kit. We do offer the PureLink HiPure Precipitator as a stand-alone product (Cat. Nos. K210021 and K210022).

Can the PureLink HiPure Precipitator be re-used?

It is not recommended. We cannot guarantee the yield or the quality of the DNA from a re-used Precipitator.

What is the minimum volume to elute my plasmid DNA when using the PureLink HiPure Precipitators?

Elution volume depends on the desired concentration. At the Midiprep scale, we typically see maximum recovery from elution volumes of 500 µL. For Maxipreps, maximum recovery was achieved at elution volumes 750 µL. Please see the product manual for the PureLink HiPure Precipitator Module.


For Research Use Only. Not for use in diagnostic procedures.