missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ HiPure BAC Buffer Kit

Catalog No. K210018
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K210018 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K210018 Supplier Invitrogen™ Supplier No. K210018

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

The PureLink HiPure BAC Buffer Kit includes Resuspension Buffer (R3) (250mL), Lysis Buffer (L7) (250mL), Precipitation Buffer (N3) (250mL), and RNase A (20 μg/mL) (5mL). All components of the PureLink HiPure BAC Buffer Kit are shipped at room temperature. Store all components at room temperature upon receipt.

PureLink HiPure BAC Buffer Kit provides additional buffers and reagents for performing BAC (Bacterial Artificial Chromosome) DNA isolation using PureLink HiPure Plasmid Kits.

  • PureLink HiPure BAC Buffer Kit provides additional buffers and reagents for performing BAC (Bacterial Artificial Chromosome) DNA isolation using PureLink HiPure Plasmid Kits
  • PureLink HiPure Plasmid Kits utilize proprietary anion-exchange resin which allows you to purify high-quality BAC DNA from E. coli cultures
  • BAC DNA requires the use of large bacterial culture volumes to obtain a reasonable DNA yield
  • From 100 mL culture, expect to purify ∽40 μg of BAC DNA
  • PureLink HiPure BAC Buffer Kit provides reagents sufficient for five BAC Maxipreps, or 25 BAC Midipreps, or 100 BAC Minipreps

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage The PureLink™ HiPure BAC Buffer Kit includes Resuspension Buffer (R3) (250 mL), Lysis Buffer (L7) (250 mL), Precipitation Buffer (N3) (250 mL), and RNase A (20 mg/mL) (5 mL). All components of the PureLink™ HiPure BAC Buffer Kit are shipped at room temperature. Store all components at room temperature upon receipt.
Format Kit
Isolation Technology Anion Exchange Resin
Sample Type Bacterial Culture
Final Product Type BAC DNA
For Use With (Application) Sequencing, Cloning, Transformation, Transfection, In Vitro Transcription
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line PureLink
Product Type BAC Buffer Kit
Quantity 1 kit
Shipping Condition Room Temperature
Target BAC DNA, Plasmid DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.