missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Genomic Plant DNA Purification Kit

Catalog No. K183001
Encompass
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K183001 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K183001 Supplier Invitrogen™ Supplier No. K183001
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The PureLink Genomic Plant DNA Purification Kit allows total DNA purification from a variety of plant samples in under one hour.

The PureLink Genomic Plant DNA Purification Kit allows total DNA purification from a variety of plant samples such as spinach, tomato, soy, wheat, Arabidopsis, mushroom, alfalfa sprouts, sunflower sprouts, corn husks, soybeans, mushroom, tomato leaves, wheat grass, and Arabidopsis thaliana leaves in under one hour.

  • Efficient—Yields high quality DNA with minimal RNA contamination
  • Simple DNA Purification from Plant Samples
  • PureLink Genomic Plant DNA Purification Kit is suitable for isolating DNA from chloroplasts and variety of plant tissues
  • Entire protocol relies on centrifugation of spin columns in absence of organic solvents
  • PureLink Genomic Plant DNA Purification Kit is designed to purify DNA from up to 100 mg of plant tissue

Maximize Yields of Total DNA

  • High yield of DNA and minimal DNA degradation is achieved by using young plant samples such as leaves and by freezing sample in liquid nitrogen immediately after collection
  • Total DNA is eluted in two aliquots of 100μL each to obtain greater DNA yield, with first elution generating between 65-80% DNA recovery and the second elution > 90%
  • Typically, DNA isolated using the PureLink Genomic Plant DNA Purification Kit has an A260/A280 > 1.80 when samples are diluted in Tris-HCl (pH 7.5), indicating that the DNA is reasonably free of proteins that could interfere with downstream applications

Isolated DNA can be used for a Number of Downstream Applications:

  • Total DNA purification from different plant tissue with the PureLink Genomic Plant DNA Purification Kit is suitable for a variety of applications, including PCR, restriction enzyme digestion, and southern blotting

Specifications

Column Type Spin Column
Content And Storage • 12.5 mL Resuspension Buffer R2; room temperature
• 5 mL Precipitation Buffer N2; room temperature
• 7.5 mL Binding Buffer B4; room temperature
• 25 mL Wash Buffer W4; room temperature
• 10 mL Wash Buffer W5; room temperature
• 10 mL Elution Buffer; room temperature
• 0.75 mL 20% SDS; room temperature
• 0.75 mL RNase A; room temperature
• 50 PureLink Spin Columns with Collection Tubes; room temperature
• 50 Collection Tubes; room temperature

Isolation Technology Silica Spin Column
Scale Mini
Sample Type Plants
Elution Volume 100 to 200 μL
Final Product Type gDNA
For Use With (Application) PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
High-throughput Compatibility Not High-throughput Compatible (Manual)
Quantity 50 Preps
Shipping Condition Room Temperature
Test Time 40 min.
Yield Up to 14.6 μg
Starting Material Amount 100 mg
Show More Show Less
I'm interested in isolating plant DNA, but am worried about the abundance of carbohydrates and complex polysaccharides in the tissue. What kit do you recommend for this?

We offer several kits specifically designed to isolate plant DNA depending on the amount of starting material and chemistry you are interested in using. Please use this link for a comparison of our kits available for plant DNA extraction.

For Research Use Only. Not for use in diagnostic procedures.