missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Genomic Lysis/Binding Buffer

Catalog No. K182302
Encompass
Change view
Click to view available options
Quantity:
80 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K182302 80 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K182302 Supplier Invitrogen™ Supplier No. K182302

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

PureLink Genomic Spin Columns, Collection Tubes, Digestion Buffer, Lysis/Binding Buffer, Wash Buffers, Elution Buffer, Proteinase K, RNase A, and the manual. The PureLink 96 Genomic DNA Kit includes PureLink Genomic Binding Plates, Wash Plates, Deep Well Plates, Foil Tape, Digestion Buffer, Lysis/Binding Buffer, Wash Buffers, Elution Buffer, Proteinase K, RNase A, and the manual.

The PureLink™ Genomic DNA Mini Kit and PureLink™ 96 Genomic DNA Kit enable high-yield, high-purity DNA extractions from wide variety of sample types, including blood, tissues, cells, bacteria, swabs, and blood spots, in familiar silica spin-column or semi-skirted 96-well plate format.

The PureLink™ Genomic DNA Mini Kit and PureLink™ 96 Genomic DNA Kit enable high-yield, high-purity DNA extractions from wide variety of sample types, including blood, tissues, cells, bacteria, swabs, and blood spots, in familiar silica spin-column or semi-skirted 96-well plate format.

  • Obtain higher gDNA yields and purity—Optimized spin-column membrane and buffer formulations
  • Save time and money—Use with large range of sample sizes (e.g., 100 μL to 1 mL blood); 96-well version does not require special equipment
  • Increase flexibility—one kit works with variety of sample types

Order Info

Shipping Condition: Room Temperature

Specifications

Buffer Lysis Buffer, Binding Buffer
Column Type Spin Column
Content And Storage Store at room temperature.
Format Kit
Sample Type Bacteria, Blood, Blood Spot, Cells, Swab, Tissue
Product Line PureLink
Product Type Genomic Lysis/Binding Buffer
Quantity 80 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.