Learn More
Invitrogen™ PureLink™ Genomic Digestion Buffer
Description
Includes
PureLink Genomic Spin Columns, Collection Tubes, Digestion Buffer, Lysis/Binding Buffer, Wash Buffers, Elution Buffer, Proteinase K, RNase A, and the manual. The PureLink 96 Genomic DNA Kit includes PureLink Genomic Binding Plates, Wash Plates, Deep Well Plates, Foil Tape, Digestion Buffer, Lysis/Binding Buffer, Wash Buffers, Elution Buffer, Proteinase K, RNase A, and the manual.
The PureLink™ Genomic DNA Mini Kit and PureLink™ 96 Genomic DNA Kit enable high-yield, high-purity DNA extractions from wide variety of sample types, including blood, tissues, cells, bacteria, swabs, and blood spots, in familiar silica spin-column or semi-skirted 96-well plate format.
- Obtain higher gDNA yields and purity—Optimized spin-column membrane and buffer formulations
- Save time and money—Use with large range of sample sizes (e.g., 100 μL to 1 mL blood); 96-well version does not require special equipment
- Increase flexibility—one kit works with variety of sample types
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Buffer | Digestion Buffer |
| Column Type | Spin Column |
| Content And Storage | Store at room temperature. |
| Format | Kit |
| Sample Type | Bacteria, Blood, Blood Spot, Cells, Swab, Tissue |
| Product Type | Genomic Digestion Buffer |
| Quantity | 70 mL |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.