Learn More
Invitrogen™ Proteinase K, recombinant
Proteinase K from the fungus Engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins.
Supplier: Invitrogen™ 25530015
Description
Proteinase K from the fungus Engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins.
- Over a wide pH range–optimal activity between 6.5 and 9.5
- Under denaturing conditions–e.g., in the presence of SDS or urea
- In the presence of metal chelating agents–e.g., EDTA
- At comparatively high temperatures–optimum digestion temperature is 65°C
- Protein Filter: P
Applications
Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.
Performance and quality testing
Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.
Unit definition
One mAnson unit is described as that amount of enzyme that liberates 1μmole of Folin-positive amino acid within one minute at 37°C using hemoglobin as a substrate.
Order Info
Approved for shipment at Room Temperature or on Wet Ice
Chemical Identifiers
| Approved for shipment at Room Temperature or on Wet Ice | |
| Proteinase K | |
| Powder |
Specifications
| One vial Store in refrigerator (2–8°C). |
|
| 100 mg | |
| Powder |
| Approved for shipment at Room Temperature or on Wet Ice | |
| Proteinase K |
For Research Use Only. Not for use in diagnostic procedures.