Learn More
Invitrogen™ SuperScript™ III Platinum™ SYBR™ Green One-Step qPCR Kit w/ROX
Combine a powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression.
$1,084.00 - $4,630.00
Specifications
| Concentration | 2X |
|---|---|
| Content And Storage | Store in freezer (-5 to -30°C). |
| Detection Method | SYBR |
| PCR Method | 1-step RT-qPCR |
| Polymerase | Taq DNA Polymerase |
| missing translation for 'productCode' | missing translation for 'brand' | No. of Reactions | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| missing translation for 'productCode' | missing translation for 'brand' | No. of Reactions | Price | Quantity & Availability | |||||
11746100
|
Invitrogen™
11746100 |
100 Reactions |
Each for $1,084.00
|
|
|||||
11746500
|
Invitrogen™
11746500 |
500 Reactions |
Each for $4,630.00
|
|
|||||
Description
One-Step qRT-PCR Kits with ROX combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient, high-throughput one-step format. SuperScript™ III Reverse Transcriptase produces higher yields of full-length cDNA for broader gene representation. Platinum™ Taq DNA Polymerase activates faster and more completely than other hot-start enzymes to deliver more sensitive amplification. ROX reference dye premixed at optimal concentration for ABI instruments.
- Provides a highly efficient, cost-effective qRT-PCR system with SYBR Green I detection
Order Info
Shipping Conditions: Dry Ice
Specifications
| 2X | |
| SYBR | |
| Taq DNA Polymerase | |
| 7000 System, 7300 System, 7700 System, 7900HT System | |
| Real Time PCR SYBR Master Mix | |
| Gene Expression |
| Store in freezer (-5 to -30°C). | |
| 1-step RT-qPCR | |
| Standard | |
| Platinum, SYBR, SuperScript | |
| Dry Ice | |
| RNA |
For Research Use Only. Not for use in diagnostic procedures.