missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ SYBR™ Green qPCR SuperMix-UDG DFS Item

Provides a sensitive and convenient system for real-time PCR detection with SYBR™ Green I dye.

Supplier:  Invitrogen™ 11733038

 View more versions of this product

Catalog No. 11733038


Only null left
Explore available promotions

Description

Description

The SuperMix format combines amplification and detection reagents into a ready-to-go mix to make set-up easy and ensure reproducible results.

  • Platinum™ Taq DNA Polymerase for sensitive amplification from as few as 10 copies of target
  • 100% dUTP and UDG enzyme for superior control of carryover contamination, saving time by reducing the number of failed experiments
  • Separate tubes of ROX reference dye and BSA for easy optimization on 96-well plates and glass capillary tube instruments
  • SYBR™ Green I dye for simple and easy detection of real-time product

Order Info

Shipping Conditions: Dry Ice

Specifications

Specifications

• Platinum™ SYBR™ Green qPCR SuperMix-UDG (2 × 1.25 mL)
• 50 mM Magnesium Chloride (1 mL)
• ROX Reference Dye (100 μL)
• 20X BSA (300 μL)

Sufficient reagents are provided for 100 reactions of 50 μL each. Store all components at -20°C.
Tube
qPCR
Standard
Platinum, SYBR
100 reactions
100 Reactions of 50 μL Each
100 Reactions
2 x 1.25 mL
SYBR
High
Taq DNA Polymerase
7000 System, 7300 System, 7500 System, 7700 System, 7900HT System, GeneAmp 5700, StepOne™, StepOnePlus™, ViiA™ 7 System, Cepheid SmartCycler, BioRad iCycler iQ
Real Time PCR SYBR Master Mix
Dry Ice
Gene Expression
DNA (Genomic), cDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.