missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ III Platinum™ One-Step qRT-PCR Kit

Combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient one-step format.

$1,150.00 - $4,600.00

Specifications

Detection Method Primer-probe
PCR Method 1-step RT-qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7500 System, BioRad iCycler iQ, BioRad iQ5, Stratagene Mx4000, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Cepheid SmartCycler, BioRad MyiQ
View More Specs

Includes: 100μL SuperScript III/Platinum Taq Mix; 2 x 1.25mL 2X Reaction Mix; 1mL Magnesium Sulfate (50mM); 100μL ROX Reference Dye (25μM)

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
11732020
Encompass
View Documents Promotion Details
Invitrogen™
11732020
100 Reactions
Each for $1,150.00
Only null left
 
11732088
Encompass
View Documents Promotion Details
Invitrogen™
11732088
500 Reactions
Each for $4,600.00
Only null left
 
Description

Description

SuperScript™ III Platinum™ One-Step qRT-PCR Kit provides sensitive, specific detection with fluorogenic primers and probes, while the SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit offers the ease and convenience of SYBR™ Green I detection.

  • The SuperScript III Platinum One-Step qRT-PCR Kit’s proven formula delivers reliable, accurate target detection when it matters most
  • A convenient, one-step format and consistent performance make it ideal for a number of gene expression applications, including infectious disease surveillance
  • The kit has been validated by several global public health organizations and has long been the choice of the US CDC for infectious disease surveillance
  • SuperScript III RT for higher temperature cDNA synthesis when addressing difficult RNA secondary structure
  • Platinum Taq DNA Polymerase with hot-start technology for improved specificity
  • LUX fluorogenic primers and dual-labeled fluorogenic probes for effective detection performance
  • Validated performance on multiple real-time instrument platforms, including rotor-based systems
  • Multiple SuperScript III Platinum One-Step qRT-PCR Kit configurations are available
  • Store components at -20°C, ROX Reference Dye must be stored in the dark

Order Info

Shipping Condition: Dry ice

Specifications

Specifications

Primer-probe
Taq DNA Polymerase
7500 System, BioRad iCycler iQ, BioRad iQ5, Stratagene Mx4000, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Cepheid SmartCycler, BioRad MyiQ
One-Step qRT-PCR Kit
Gene Expression, Pathogen Detection, Virus Detection
1-step RT-qPCR
Standard
Platinum, SuperScript
Dry Ice
RNA
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.