missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Platinum™ Quantitative PCR SuperMix-UDG w/ROX
qPCR SuperMixes with ROX for ABI instruments are ready-to-use reaction mixes for high-specificity, real-time amplification of cDNA, genomic DNA or plasmid DNA.
$894.00 - $3,470.00
Specifications
| Concentration | 2X |
|---|---|
| Detection Method | Primer-probe |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Polymerase | Platinum Taq DNA Polymerase |
Description
qPCR SuperMixes with ROX are available in two formats:- Platinum™ SYBR™ Green qPCR SuperMix-UDG with ROX provides a sensitive, cost-effective system for real-time PCR detection with SYBR™ Green I dye
- Platinum™ qPCR SuperMix-UDG with ROX offers superior performance with either fluorogenic primers (e.g., LUX™ Primers) or probe-based detection systems (e.g., TaqMan™)
Order Info
Guaranteed stable for 6 months when properly stored. Shipping Conditions: Dry Ice.
Specifications
| 2X | |
| High | |
| Platinum Taq DNA Polymerase | |
| 7000 System, 7300 System, 7700 System, 7900HT System | |
| Quantitative PCR SuperMix-UDG | |
| Gene Expression | |
| DNA (Genomic), cDNA |
| Primer-probe | |
| qPCR | |
| Standard | |
| Platinum | |
| Dry Ice | |
| 2 X |
For Research Use Only. Not for use in diagnostic procedures.