Learn More
Invitrogen™ Dynabeads™ Streptavidin Magnetic Beads
Description
Includes
Supplied in PBS, pH 7.4 with 0.1% BSA and 0.02% NaN3 added as preservatives.
Dynabeads Streptavidin Magnetic Beads are designed for high-specificity, low-background capture of target molecules with biotinylated affinity binders such as antibodies, antigens, or oligonucleotides. Available in various sizes and surface chemistries, these beads can be tailored to provide the best fit for specific application needs.
Dynabeads Streptavidin Magnetic Beads are compatible with high-throughput automation platforms, including KingFisher sample purification systems, ensuring seamless integration into new and existing workflows.
Key advantages of Dynabeads Streptavidin Magnetic Beads
- ISO 13485–certified manufacturing—exceptional manufacturing control to ensure robust and consistent performance between production lots, meeting international quality standards for medical devices
- Monosized, monodispersed magnetic beads—uniform particle size provides consistent and reproducible results across applications
- Dual size options—available in 1-μm and 2.8-μm diameter specifications to accommodate diverse experimental requirements and application-specific needs
- Multiple surface chemistry options—various surface functionalization chemistries available to optimize binding efficiency and compatibility for specific applications
- Application-optimized design—size and surface chemistry combinations enable best-fit selection for targeted experimental protocols and assay requirements
Precision-engineered manufacturing delivers exceptional control over critical bead properties
Dynabeads Streptavidin products are based on magnetic polystyrene-based beads that are covalently coupled with recombinant streptavidin, creating a robust platform for target capture and isolation using biotinylated affinity binders. Dynabeads Streptavidin product manufacturing enables exceptional precision over three critical parameters: size distribution, magnetization properties, and surface chemical characteristics. This ensures consistent performance across batches and applications, providing the reliability necessary for reproducible experimental outcomes.
Optimized size selection enhances application-specific performance
The Dynabeads Streptavidin product line offers two distinct size options, each engineered to excel in specific experimental contexts. The 1-μm beads, available in the Dynabeads MyOne series, are ideally suited for applications requiring fast reaction kinetics, slow sedimentation rates, and high sensitivity-to-capacity ratios. These smaller beads provide enhanced surface area contact and reduced gravitational settling, making them particularly valuable for sensitive detection assays.
In contrast, the 2.8-μm beads, found in the Dynabeads M-270 and M-280 series, are perfect for applications demanding low background binding and reduced risk of aggregation in aggregation-prone applications. These larger beads offer even easier handling and faster magnetic separation compared to the Dynabeads MyOne series, facilitating streamlined workflow processing.
Tailored surface chemistry options address diverse experimental requirements
The Dynabeads Streptavidin product line includes two distinct surface chemistry configurations designed to optimize performance across varied experimental conditions. Hydrophobic surfaces are available in Dynabeads MyOne Streptavidin T1, Dynabeads Streptavidin for Target Enrichment, and Dynabeads M-280 Streptavidin products. These surfaces are BSA-blocked after streptavidin coupling making them ideal for high signal-to-noise applications such as immunoassays, target enrichment, and on-bead PCR for next-generation sequencing.
Alternatively, hydrophilic surfaces are featured in Dynabeads MyOne Streptavidin C1, Dynabeads Streptavidin for In Vitro Transcription, and Dynabeads M-270 Streptavidin products. These surfaces contain no BSA blocking and are particularly suitable for hydrophobic targets, harsh lysis conditions, sample preparation workflows for mass spectrometry, and applications demanding low or no nuclease activity, such as on-bead reverse transcription or in vitro transcription.
For researchers uncertain about which bead type best suits their application, a Dynabeads Streptavidin Trial Kit is available. It allows for side-by-side performance evaluation of different bead formulations under your specific experimental conditions, ensuring optimal selection for your research needs.
ISO 13485 certification ensures scalable quality and OEM support
Manufactured under ISO 13485 certification, Dynabeads Streptavidin products deliver outstanding batch-to-batch consistency. This robust quality assurance framework ensures that once customers develop an assay or workflow, they can rely on reproducible results across multiple experimental iterations. This consistency makes these versatile beads an efficient solution for applications spanning the entire spectrum from fundamental research to commercial manufacturing processes. For researchers developing commercial products or services, or those requiring larger bulk volumes, specialized support is available by emailing oemdynal@thermofisher.com.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Bead Type | Magnetic polystyrene-based beads covalently coupled with recombinant Streptavidin |
| Concentration | 10 mg/mL |
| Content And Storage | Dynabeads MyOne Streptavidin T1 are supplied in PBS, pH 7.4, with 0.1% BSA and 0.02% NaN3 . Store at 2–8°C. |
| Format | Beads in Suspension |
| Isolation Technology | Magnetic Bead |
| Diameter (Metric) | 1 μm (CV < 5%) |
| For Use With (Equipment) | KingFisher™ Sample Purification System, DynaMag™ magnets |
| For Use With (Application) | Immunoassays, IP, ChIP, RIP, cell isolations, or DNA isolation for downstream amplification reactions or NGS, requiring high sensitivity, rapid target capture with compatibility with subsequent enzymatic reactions |
| High-throughput Compatibility | High-throughput Compatible |
| Product Line | Dynabeads |
| Show More |
Frequently Asked Questions (FAQs)
We do not recommend this as streptavidin becomes hydrophobic and aggregates during denaturation.
Please review the following possibilities for why your Dynabeads magnetic beads are not pelleting:
- The solution is too viscous.
- The beads have formed aggregates because of protein-protein interaction.
Try these suggestions:
- Increase separation time (leave tub on magnet for 2-5 minutes)
- Add DNase I to the lysate (~0.01 mg/mL)
- Increase the Tween 20 concentration to ~0.05% of the binding and/or washing buffer.
- Add up to 20 mM beta-merecaptoethanol to the binding and/or wash buffers.
Which product to choose depends on the properties of your sample, the buffers and solutions applied, as well as the downstream application. In general, all Dynabeads Streptavidin beads can be used in applications involving biotinylated ligands; however, some beads may perform better than others in particular applications due to their characteristics. Dynabeads M-280 Streptavidin beads and Dynabeads MyOne Streptavidin T1 beads are commonly used for protein and nucleic acid applications. Dynabeads M-270 Streptavidin beads and MyOne Streptavidin C1 beads are preferred for nucleic acid diagnostics, specifically with samples that have a high concentration of chaotropic salts, immunoassays involving small biotinylated antigens, and in applications that are not compatible with BSA, as these beads are not blocked with BSA. Dynabeads MyOne Streptavidin beads offer increased binding capacity and slower sedimentation rate, making them ideal for automated applications and when larger amounts of a biotinylated compound or its specific target need to be isolated. Please see the selection guide here ( https://www.thermofisher.com/us/en/home/brands/product-brand/dynal/streptavidin-coupled-dynabeads.html?icid=fr-strep-1).
You can assay the supernatant for unbound nucleic acid to determine the amount of nucleic acid bound to the Dynabeads Streptavidin beads. The concentration of nucleic acids can be checked by measuring the OD or by running them on a gel. Alternatively, the nucleic acids can be labeled radioactively and the concentration measured directly on the beads, or fluorescently and measured in the supernatant.
Our Dynabeads Streptavidin magnetic beads can be used directly in PCR or real-time PCR. However, you would have to empirically optimize the amount of beads to be used per volume of reaction.
For Research Use Only.