missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ SILANE Viral NA Kit

Catalog No. 37011D
Encompass
Change view
Click to view available options
Quantity:
96 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37011D 96 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37011D Supplier Invitrogen™ Supplier No. 37011D

Includes

Dynabeads MyOne; SILANE (5mL, 40mg/mL, see Cat. no. 370-02D for further characteristics), binding/washing/elution buffers and a protocol optimized for isolation of viral DNA/RNA. The kit content is sufficient for 96 isolations.

For sensitive, highly predictable and consistent extraction and isolation of viral DNA and RNA from human serum/plasma samples, following a simple separation protocol.

For sensitive, highly predictable and consistent extraction and isolation of viral DNA and RNA from human serum/plasma samples, following a simple separation protocol. The 1 μm Dynabeads have an optimized silica-like surface chemistry, and a high specific surface area, providing efficient kinetics and a high sensitivity. Their uniform size and surface area ensure reproducible results.

A higher sensitivity and improved performance is achieved with Dynabeads™ compared to magnetic particles from alternative suppliers. Other particles often have a random size range distribution, surface area and binding capacity. This could compromise the reproducibility of your results. Dynabeads™ hold reputable Dynal high standards with respect to within- and between-lot reproducibility and automation ability.

Advantages
• Isolates viral DNA and viral RNA
• High sensitivity, allowing for a low detection limit
• Effective isolation of nucleic acids even from challenging viruses (e.g. HBV)
• Can detect HBV DNA in a 200 μl serum sample containing only 40 infectious units (using 2 mg (50 μl) Dynabeads™)
• Quicker than spin-columns
• Cost-effective and performs to the same level as spin-columns
• Automation-friendly (slow sedimentation rate + high magnetic mobility)

Applications
• Isolation of viral nucleic acids (DNA/RNA)
• From human serum/plasma samples
• Well suited for automated assays
• Separate kit available for the isolation of genomic DNA (Cat. no. 370-12D)

Additional Info
Validation and customization (bead, buffer, protocol or format) can be made available on a custom OEM basis. For further information, please contact us at: ivd@invitrogen.com

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage

The kit contains: Dynabeads™ MyOne™ SILANE (5 mL, 40 mg/mL, see Cat. No. 370-02D for further characteristics), binding/washing/elution buffers and a protocol optimized for isolation of viral DNA/RNA. The kit content is sufficient for 96 isolations.

Store at: 2°C to 8°C.

Isolation Technology Magnetic Bead
Sample Type Plasma, Serum
Elution Volume 100 μL
Final Product Type Viral DNA and RNA
For Use With (Equipment) Automated Liquid Handling Systems
For Use With (Application) RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern/Southern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling, hybridization
High-throughput Compatibility High-throughput Compatible
Quantity 96 preps
Shipping Condition Room Temperature
Size 1 μm
Target Viral DNA/RNA
Test Time Up to 45 min.
Starting Material Amount Serum: 200 μL
Plasma: 200 μL
Show More Show Less
My Dynabeads magnetic beads are not pelleting well with the magnet. Do you have any suggestions for me?

Please review the following possibilities for why your Dynabeads magnetic beads are not pelleting:

- The solution is too viscous.
- The beads have formed aggregates because of protein-protein interaction.

Try these suggestions: - Increase separation time (leave tub on magnet for 2-5 minutes)
- Add DNase I to the lysate (~0.01 mg/mL)
- Increase the Tween 20 concentration to ~0.05% of the binding and/or washing buffer.
- Add up to 20 mM beta-merecaptoethanol to the binding and/or wash buffers.

What is the principle behind isolation of viral nucleic acids with the Dynabeads SILANE Viral NA Kit?

The Dynabeads SILANE Viral NA Kit will bind genomic DNA as well as viral nucleic acids, but the buffers have been optimized for stronger binding of shorter fragments and very small amounts of viral DNA/RNA. This buffer is not advised for genomic DNA binding, because the interaction with the bead surface will be stronger, which will require more work to elute genomic DNA than viral nucleic acids.

Can Dynabeads SILANE beads be used for nucleosome isolation?

That depends. If you want to isolate intact nucleosomes (histones + DNA complex), that is not possible with Dynabeads SILANE beads. But you can isolate DNA from nucleosomes if they are not cross-linked. For chromatin immunoprecipitation, you could consider the MAGnify Chromatin Immunoprecipitation System.

How much DNA is recovered per isolation using the Dynabeads DNA DIRECT Universal Kit?

The yield will depend on the number of nucleated cells present in the sample. One unit (200 µL) of Dynabeads DNA DIRECT Universal Kit will isolate at least 200 ng of high quality, PCR-ready genomic DNA per sample, sufficient for 10 PCR reactions. The size of the sample used should be adjusted to match the Dynabeads magnetic beads capacity. A typical starting sample is 10,000 cultured cells or 10 µL blood. The protocol can be adjusted to support 30 µL blood samples, sufficient for 30-50 downstream PCR amplifications (600 ng to 1 µg DNA).

I have a long double-stranded DNA fragment I would like to isolate. What product do you recommend?

For biotin-labled DNA that is less than 1 kb, we recommend you use Dynabeads M270 Streptavidin and MyOne C1 magnetic beads. We recommend our Dynabeads KilobaseBINDER Kit, which is designed to immobilize long (>1 kb) double-stranded DNA molecules. The KilobaseBINDER reagent consists of M-280 Streptavidin-coupled Dynabeads magnetic beads along with a patented immobilization activator in the binding solution to bind to long, biotinylated DNA molecules for isolation. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/immobilisation-of-long-biotinylated-dna-fragments.html) for more information in regards to long biotinylated DNA fragment isolation.

Can I use Dynabeads magnetic beads to isolate single-stranded DNA templates?

Yes, Dynabeads magnetic beads can be used to isolate single-stranded DNA. Streptavidin Dynabeads magnetic beads can be used to target biotinylated DNA fragments, followed by denaturation of the double-stranded DNA and removal of the non-biotinylated strand. The streptavidin-coupled Dynabeads magnetic beads will not inhibit any enzymatic activity. This enables further handling and manipulation of the bead-bound DNA directly on the solid phase. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/preparing-single-stranded-dna-templates.html) for more information in regards to single-stranded DNA capture.

What is the magnetic susceptibility for Dynabeads magnetic beads?

Magnetic susceptibility is a measure of how quickly the beads will migrate to the magnet. This will depend on the iron content and the character of the iron oxide. The magnetic susceptibility given for the Dynabeads magnetic beads is the mass susceptibility, given either as cgs units/g or m^3/kg (the latter being an SI unit). For ferri- and ferromagnetic substances, the magnetic mass susceptibility is dependent upon the magnetic field strength (H), as the magnetization of such substances is not a linear function of H but approaches a saturation value with increasing field. For that reason, the magnetic mass susceptibility of the Dynabeads magnetic beads is determined by a standardized procedure under fixed conditions. The magnetic mass susceptibility given in our catalog is thus the SI unit. Conversion from Gaussian (cgs, emu) units into SI units for magnetic mass susceptibility is achieved by multiplying the Gaussian factor (emu/g or cgs/g) by 4 pi x 10^-3. The resulting unit is also called the rationalized magnetic mass susceptibility, which should be distinguished from the (SI) dimensionless magnetic susceptibility unit. In general, magnetic mass susceptibility is a measure of the force (Fz) influencing an object positioned in a nonhomogenous magnetic field. The magnetic mass susceptibility of the Dynabeads magnetic beads is measured by weighing a sample, and then subjecting the sample to a magnetic field of known strength. The weight (F1) is then measured, and compared to the weight of the sample when the magnetic field is turned off (F0). The susceptibility is then calculated as K x 10^-3 = [(F1-F0) x m x 0.335 x 10^6], where K is the mass susceptibility of the sample of mass m. The susceptibility is then converted to SI units.

How can I determine coupling efficiency of Dynabeads magnetic beads?

There are different methods to check binding of ligands to the beads, including optical density (OD) measurement, fluorescent labeling, and radioactive labeling.

For OD measurement, you would measure the OD of the ligand before immobilization to the beads and compare it with the ligand concentration that is left in the supernatant after coating. This gives a crude measurement of how much protein has bound to the beads.

Protocol:

1.Set spectrophotometer to the right wavelength. As a blank, use the Coupling Buffer.
2.Measure the absorbance of the Pre-Coupling Solution. A further dilution may be necessary to read the absorbance, depending upon the amount of ligand added.
3.Measure the absorbance of the Post-Coupling Solution. A dilution may be necessary to read the absorbance.
4.Calculate the coupling efficiency, expressed as the % protein uptake, as follows. [(Pre-Coupling Solution x D) - (Post-Coupling Solution x D)] x 100/(Pre-Coupling Solution x D) where D = dilution factor.

For fluorescent labeling, we suggest negatively quantifying the amount of ligand bound by measuring ligand remaining in the coupling supernatant (compared to the original sample), rather than directly measuring the ligands on the beads. Add labeled ligand to the beads, and measure how much ligand is left in the supernatant (not bound to the beads). By comparing this with the total amount added in the first place, you can then calculate how much of the ligand that has been bound to the beads. Keep in mind that the Dynabeads magnetic beads are also autofluorescent, which is why direct measuring of fluorescence of the bead-bound ligands is not recommended, but rather this indirect approach. The label could be, for example, FITC/PE. Some researchers perform a direct approach with success (using a flow cytometer).

Radioactive labeling is the most sensitive method of the three, but it is also the most difficult one. It involves radioactively labeling a portion of the ligand. We use radiolabeled I-125 in tracer amounts and mix it with "cold" ligands in a known ratio before coupling. The absolute quantities for the ligand on the beads should be obtained by measuring the beads in a scintillation (gamma) counter and comparing the cpm with a standard.

Protocol:

1.Take out an appropriate amount of beads and wash the beads in 1 mL of binding buffer.
2.Pipette out desired amount of human IgG in a separate tube.
3.Mix the human IgG with I-125-labeled human IgG (30,000 - 100,000 cpm).
4.Dilute the mixture of human IgG and I-125-labeled human IgG to 100 mL in binding buffer.
5.Incubate for 30 minutes at room temperature and measure the cpm in a scintillation counter.
6.Wash the beads (with coating) four times, and measure cpm again.
The % binding is calculated by using the equation : (cpm after washing/cpm before washing)x100%.

What sizes do you offer for the Dynabeads magnetic beads?

Dynabeads magnetic beads come in three sizes: 4.5 µm (M-450), 2.8 µm (M-270/M-280), and 1 µm (MyOne beads). The largest of the Dynabeads magnetic beads is ideal for big targets like cells. The 2.8 µm beads are recommended for proteomics and molecular applications. The smallest of the beads, 1 µm, are ideal for automated handling.

Can Dynabeads magnetic beads be sonicated?

In general, short sonication is a good way to reduce aggregation of the beads and ensure optimal homogenous conditions at the time of ligand addition when coating the beads. When target is bound to the beads, more care is needed, as the binding might break. The streptavidin beads themselves should tolerate sonication. We have not tested sonication for long periods, but 5 minutes is fine. We do not have information about the streptavidin-biotin interaction being broken by such treatment.

Can Dynabeads magnetic beads be sterilized?

If desired, the uncoated epoxy or tosylactivated beads can be sterilized by washing with 70% ethanol. Coated beads cannot be sterilized.

What are Dynabeads magnetic beads?

Dynabeads magnetic beads are uniform, non-porous, superparamagnetic, monodispersed and highly cross-linked polystyrene microspheres consisting of an even dispersion of magnetic material throughout the bead. The magnetic material within the Dynabeads magnetic beads consists of a mixture of maghemite (gamma-Fe2O3) and magnetite (Fe3O4). The iron content (Fe) of the beads is 12% by weight in Dynabeads magnetic beads M-280 and 20% by weight in Dynabeads magnetic beads M-450. The Dynabeads magnetic beads are coated with a thin polystyrene shell which encases the magnetic material, and prevents any leakage from the beads or trapping of ligands in the bead interior. The shell also protects the target from exposure to iron while providing a defined surface area for the adsorption or coupling of various molecules.

Uniformity of bead size and shape provides consistent physical and chemical properties. These uniform physical characteristics lead to high-quality, reproducible results.

The Dynabeads magnetic beads are available in three different sizes: 4.5 µm (M-450 beads), 2.8 µm (M-270/M-280 beads) and 1 µm (MyOne beads). as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

How can I use Dynabeads MyOne SILANE magnetic beads for isolation of total RNA?

Please see the example protocol for isolation of total RNA from cultured cells (with enzyme):

Add 20 µl (20 mg/ml) Proteinase K prepared in Elution Buffer to 1 million freshly harvested cultured cells delivered in a volume of 50 µl. Mix and incubate at room temperature for 2 minutes.
Add 350 µl of Lysis/Binding Buffer and mix by pipetting. Incubate at room temperature for 10 minutes.
Note: Do not add Proteinase K directly to Lysis/Binding Buffer without cells .
Note: The vial containing Dynabeads MyOne SILANE magnetic beads should be resuspended (e.g. vortex) to a homogenous suspension prior to use. Leave on a roller until use.
Add 50 µl Dynabeads MyOne SILANE suspension (40 mg/ml) to the mixture.
Add 400 µl of isopropanol. Incubate on a roller at room temperature for 5 minutes.
Place the tube on a magnet and let the Dynabeads magnetic beads collect at the magnet for 3 minutes (or until the supernatant is clear). Remove the supernatant completely.
Remove the tube from the magnet and add 500 µl Washing Buffer 1. To wash, resuspend the Dynabeads magnetic beads thoroughly by pipetting.
Place the tube on a magnet and let the Dynabeads magnetic beads collect at the magnet for 2 minutes (or until the supernatant is clear). Remove the supernatant completely.
Remove the tube from the magnet and add 500 µl Washing Buffer 2. To wash, resuspend the Dynabeads magnetic beads thoroughly by pipetting.
Place the tube on a magnet and let the Dynabeads magnetic beads collect at the magnet for 2 minutes (or until the supernatant is clear). Remove the supernatant completely.
Let the pellet air-dry while still placed on the magnet (approx. 10 minutes).
Remove the tube from the magnet and add 100 µl Elution Buffer. Pipette until the pellet is completely resuspended. Incubate at room temperature for 2 minutes.
Place the tube on a magnet and let the Dynabeads magnetic beads collect at the magnet for 2 minutes (or until the supernatant is clear). Transfer the supernatant/eluate containing the purified total RNA to a fresh tube.
Add 10 µl 10x DNase Buffer to the eluates. Add 20 U DNase. Incubate at 37 degrees C for 10 minutes.

What are Dynabeads SILANE Viral NA magnetic beads?

Dynabeads SILANE Viral NA magnetic beads offer an excellent tool for highly predictable and consistent extraction and isolation of viral DNA and RNA from human serum/plasma samples, following a simple magnetic separation protocol. Dynabeads SILANE Viral NA beads exhibit high sensitivity and high capacity and their slow sedimentation rate and high magnetic mobility make them ideal for automated protocols.

What do the designations M-280, M-270, and MyOne mean on Dynabeads magnetic beads?

The M stands for magnetic. M-280 refers to hydrophobic 2.8 micron beads, while M-270 refers to hydrophilic 2.8 micron beads. MyOne refers to 1 micron beads. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

What is the detection limit when using Dynabeads magnetic beads for immunoprecipitation (IP)?

Answering this question is not straightforward. It will depend on the detection method. When using HRP (horseradish peroxidase)-based detection system or radioactivity in combination with a good antibody, very little target is required. More target is required when using an AP (alkaline phosphatase)-based detection system. When a sensitive detection system is used, detection will most likely be in the nanogram range. In some cases, pictograms of target can be detected.

What is the elution volume when using Dynabeads magnetic beads for immunoprecipitation (IP)?

Within practical limits, the elution volume can be scaled up or down to suit your experiment. However, volumes less than 10 µL become more difficult to work with. In addition, the amount of target is important. If you have a lot of beads with a lot of bound target in a small elution volume, your elution may not be very efficient. Typically, 15-100 µL of beads may be eluted in 30 µL. For efficient recovery of the antigen and/or binding partners, the elution volume should at minimum equal the volume of the beads.

How can I quantify the amount of antibody bound to Dynabeads magnetic beads?

There are several methods to quantify the amount of antibody bound to the beads. The crudest method is to measure the concentration of antibody in the coupling reaction before and after antibody attachment. Either fluorescence measurements or absorbance at 280 nm can be used. Alternatively, you could measure the amount of antibody bound to the beads by fluorescence, chemiluminescence, or radiolabeling detection methods. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

How long should I incubate my antibody with the lysates?

Incubation time will depend on the immunogenicity of the primary antibody and its binding affinity with the specific antigens. For a good primary antibody, 30-40 minutes incubation should work well. If you are working with a poor antibody or a very low-abundance protein, you could try to increase binding by incubating overnight. However, this also increases the chance of background protein binding.

When should I covalently bind the antibody to the Dynabeads surface?

If the target protein has the same molecular weight as the heavy or light chain antibody, we would recommend covalently binding the antibody to the bead surface. This can be done by either crosslinking the antibody to the Dynabeads Protein A or G magnetic beads, or secondary coated beads, or by using one of the surface-activated Dynabeads magnetic beads. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

What are the general advantages of using Dynabeads magnetic beads for protein isolation?

Using Dynabeads magnetic beads for protein isolation provides several advantages:

-Rapid binding kinetics: since the number of beads per volume for Dynabeads is approximately 1,000 times higher than for the same volume of a Sepharose slurry, the probability for Dynabeads magnetic beads to hit the target is far greater.

-Incubation time: due to the rapid binding kinetics, the protocol is usually very short.
-Low background: due to the rapid binding kinetics and the short incubation time, the background is also very low.
-Trapping of impurities: the beads offer no internal volume for binding or trapping of impurities.
-Low antibody consumption: this is because Dynabeads magnetic beads are nonporous, uniform superparamagnetic, monodispersed, highly crosslinked polystyrene microspheres consisting of an even dispersion of magnetic material throughout the bead. The beads are coated with a thin layer of a highly crosslinked polystyrene shell that encases the magnetic material and prevents any leakage from the beads or trapping of ligands in the bead interior. The outer layer also provides a defined surface area for the adsorption or coupling of various molecules such as antibodies. Uniformity of bead size and shape provide consistent physical and chemical properties. These uniform physical characteristics lead to high-quality, reproducible results.
-Reproducibility: due to easier practical handling, such as pipetting. No centrifugation steps or preclearing are required.

Are Dynabeads magnetic beads compatible with dithionite, DTT, and EDTA?

No. Not only is dithionite a reducing agent, but the strong affinity of the dithionite ion for bivalent and trivalent metal cations (M2+, M3+) allows it to enhance the solubility of iron, making it a chelating agent. As a result, the iron in the Dynabeads magnetic beads is reduced and pulled out when they are exposed to dithionite. The same is observed if Dynabeads magnetic beads are exposed to DTT and EDTA. With EDTA, we highly recommend checking the minimal amount of EDTA that your specific molecules would tolerate for binding to the Dynabeads, and if it will affect your specific application. For some applications, low concentrations of EDTA can be tolerated by Dynabeads. On the other hand, using 10 mM EDTA with heating affects the binding of biotin molecules to Dynabeads streptavidin. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

Are Dynabeads magnetic beads compatible with Urea?

Yes, they are compatible with 6-8 M Urea when used during post-coupling steps.

Are Dynabeads magnetic beads compatible with centrifugation?

Dynabeads magnetic beads, being magnetic in nature are really not designed to be centrifuged. That being said, the beads themselves are compact, as the pores in the polymer matrix are filled with magnetic material and coated with a final outer polymer shell that will further add to the rigidity of the beads. Hence, pressure should theoretically not be a problem for the beads themselves, but the force exerted by the beads on surrounding cells in the pellet may be detrimental to the cells. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

What are the benefits of using magnetic beads in immunoprecipitation (IP) experiments?

Magnetic beads, unlike agarose beads, are solid and spherical, and antibody binding is limited to the surface of each bead. While magnetic beads do not have the advantage of a porous center to increase the binding capacity, they are significantly smaller than agarose beads (1 to 4 µm), which collectively gives them adequate surface area-to-volume ratios for optimum antibody binding.

High-power magnets are used to localize magnetic beads to the side of the incubation tube and out of the way to enable cell lysate aspiration without the risk of also aspirating immune complexes bound to the beads. Magnetic separation avoids centrifugation, which can break weak antibody-antigen binding and cause loss of target protein.

However, an issue with the use of magnetic beads is that bead size variations may prevent all beads from localizing to the magnet. Additionally, while immunoprecipitation using agarose beads only requires standard laboratory equipment, the use of magnetic beads for immunoprecipitation applications requires high-power magnetic equipment that can be cost-prohibitive. Read more about our Magnetic Immunoprecipitation Products (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/immunoprecipitation.html#products).


For Research Use Only. Not for use in diagnostic procedures.