missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ λ DNA (cIind 1 ts857 Sam 7)

Catalog No. 25250010
Encompass
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25250010 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25250010 Supplier Invitrogen™ Supplier No. 25250010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Supplied at 0.35 to 0.6μg/μL in 10mM Tris-HCl (pH 7.4), 5mM NaCl, 0.1mM EDTA.

λ DNA (cIind 1 ts857 Sam 7) is isolated from a dam+ dcm+ E coli LE597 lysogen

Suitable for use as a substrate for generating molecular size standards and for testing restriction endonuclease activity. Restriction endonucleases sensitive to dam or dcm methylation will not cleave this DNA.

Performance and Quality Testing: Purity, DNA structure, and selected restriction endonuclease fragmentation patterns are verified by agarose gel analysis.

Agarose Gel Electrophoresis, Cloning, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting, Restriction Enzyme Cloning

Order Info

Shipping Condition: Wet Ice

Specifications

Product Type λ DNA
Includes Control Template(s)
Content And Storage Supplied at 0.25 to 0.6 μg/μl in 10 mM Tris-HCl (pH 7.4), 5 mM NaCl, 0.1 mM EDTA. Store at +4°C.
Label or Dye Unconjugated
Shipping Condition Wet Ice
Format Tube
Quantity 500 μg

For Research Use Only. Not for use in diagnostic procedures.