missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gateway™ BP Clonase™ Enzyme Mix

$1,606.00 - $6,580.00

Specifications

Shipping Condition Dry Ice
Enzyme BP Clonase
Compatible Buffer Reaction Buffer
Product Type BP Clonase Enzyme Mix
Product Line Clonase, Gateway
View More Specs

Includes: Proteinase K solution (2μg/μL), 30% PEG 8000/30mM MgCl2 Solution and a positive control vector.

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11789013
Encompass
View Documents Promotion Details
Invitrogen™
11789013
20 Reactions
Each for $1,606.00
Only null left
 
11789021
Encompass
View Documents Promotion Details
Invitrogen™
11789021
100 Reactions
Each for $6,580.00
Only null left
 
Description

Description

  • Proteins catalyze in vitro recombination of PCR products or DNA segments from clones (containing attB sites) and Donor vector (containing attP sites) to generate Entry clones
  • BP Clonase II enzyme format is single mix of both enzyme and buffer to ensure enzyme stability and enable convenient 10μL reaction set up with fewer pipetting steps, which both maximize recombination efficiency and minimize pippetting
  • Original format of BP Clonase enzyme is stand alone enzyme blend and reaction buffer provided in separate tubes

Cloning, Gateway Cloning, Gateway Recombination

Order Info

Shipping Conditions: Dry ice

Specifications

Specifications

Dry Ice
Reaction Buffer
Clonase, Gateway
BP Clonase
BP Clonase Enzyme Mix
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.