Learn More
Invitrogen™ ChargeSwitch™ Wash Buffer W12
Description
Wash buffer used in all ChargeSwitch kits including the ChargeSwitch magnetic beads and ChargeSwitch-Pro column kits.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Buffer | Wash Buffer |
| Content And Storage | Store at room temperature. |
| Product Line | ChargeSwitch |
| Product Type | Wash Buffer W12 |
| Quantity | 1 L |
| Shipping Condition | Room Temperature |
Frequently Asked Questions (FAQs)
We strongly recommend using the Elution Buffer provided in the kit, and do not recommend elution with water. If you need to elute in any other buffer, be sure to use a buffer of pH 8.5-9.0 for efficient DNA elution.
The filter kit is based upon a dual nested column design, which facilitates the fastest and simplest protocol for obtaining high quality plasmid (less than 10 EU/µg DNA). Modified wash buffer 1 removes more endotoxin.
The typical yield is 25 µg from a 5 mL culture for the ChargeSwitch Pro mini kit, 200 µg from a 25 mL culture for the midi kit, and 800 µg from a 100 mL culture from the maxi kit. The plasmid size range for the kits is typically 3-9 kb.
Based on a patented switchable chemistry, ChargeSwitch purification utilizes the charge on a solid surface for nucleic acid capture. At lower pH, the surface chemistry holds a positive charge and attracts nucleic acid. At higher pH, the surface chemistry is neutral and any bound nucleic acid is released.
For Research Use Only. Not for use in diagnostic procedures.