missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™-Pro PCR Clean-Up Kit
GREENER_CHOICE

Catalog No. CS32250
Encompass
Change view
Click to view available options
No. of Reactions:
250 Preps
50 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
CS32250 250 Preps
CS32050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CS32250 Supplier Invitrogen™ Supplier No. CS32250
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Kit contains columns, collection tubes, recovery tubes, binding buffer, wash buffer, and elution buffer.

The ChargeSwitch™-Pro PCR Clean-up Kit purifies high-quality DNA fragments without the use of ethanol or harsh salts known to interfere with enzymatic reactions.

The ChargeSwitch™-Pro PCR Clean-up Kit purifies high-quality DNA fragments without the use of ethanol or harsh salts known to interfere with enzymatic reactions.

• 100% ethanol-free, guanidinium-free, organic solvent-free protocols
• Improved downstream performance
• Complete kit for fast, hassle-free setup
• 20% fewer handling steps than most PCR clean-up protocols
• Purification of up to 500 μL of PCR product in one column

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage

The ChargeSwitch™-Pro PCR Clean-up Kit contains columns, collection tubes, recovery tubes, binding buffer, wash buffer, and elution buffer.

Store at room temperature. Guaranteed stable for 6 months when properly stored.

Format Kit
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line ChargeSwitch
Product Type PCR Clean-Up Kit
Quantity 250 Preps
Shipping Condition Room Temperature
Target DNA from Enzymatic Reactions (e.g. PCR)
For Use With (Application) Next-Generation Sequencing (NGS), Cloning, Standard PCR
Isolation Technology ChargeSwitch™ Chemistry
No. of Reactions 250 Preps
Sample Type PCR Products
Show More Show Less
I used water to elute my plasmid instead of the Elution Buffer with my ChargeSwitch kit, but saw very low plasmid yield. Why is this?

We strongly recommend using the Elution Buffer provided in the kit, and do not recommend elution with water. If you need to elute in any other buffer, be sure to use a buffer of pH 8.5-9.0 for efficient DNA elution.

What is the difference between the ChargeSwitch Pro kit and the Charge-Switch Pro filter kit?

The filter kit is based upon a dual nested column design, which facilitates the fastest and simplest protocol for obtaining high quality plasmid (less than 10 EU/µg DNA). Modified wash buffer 1 removes more endotoxin.

What is the typical yield when using the ChargeSwitch Pro Filter Plasmid Mini or Midi Kits?

The typical yield is 25 µg from a 5 mL culture for the ChargeSwitch Pro mini kit, 200 µg from a 25 mL culture for the midi kit, and 800 µg from a 100 mL culture from the maxi kit. The plasmid size range for the kits is typically 3-9 kb.

I'm seeing bead carryover with the ChargeSwitch PCR Clean-up Kit. Is this okay for my PCR?

ChargeSwitch coated beads are inert and will not affect PCR. We have found that volumes of as much as 10 µL of beads have no deleterious effects on regular PCR when spiked in. Excessive quantities greater than 10 µL of beads will start to inhibit PCR. Some specific applications may be affected by beads, including real-time reactions or MALDI-TOF. In these applications, repeat the final elution binding as described in the protocol.

How does ChargeSwitch technology work?

Based on a patented switchable chemistry, ChargeSwitch purification utilizes the charge on a solid surface for nucleic acid capture. At lower pH, the surface chemistry holds a positive charge and attracts nucleic acid. At higher pH, the surface chemistry is neutral and any bound nucleic acid is released.

For Research Use Only. Not for use in diagnostic procedures.