Learn More
Invitrogen™ ChargeSwitch™ Lysis Buffer L13

Description
- ChargeSwitch Forensic DNA Purification Kit is magnetic bead method designed to accommodate all forensic crime scene sample types for STR profiling
- Scalable for single-tube use or for automated extraction
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Buffer | Lysis Buffer |
| Content And Storage | All components of the ChargeSwitch™ Forensic DNA Purification Kits are shipped at room temperature. Upon receipt, store the Proteinase K at 4°C. Store all other components at room temperature. All components are guaranteed stable for 6 months, if stored properly. |
| Format | Kit |
| For Use With (Application) | DNA Extraction |
| Product Line | ChargeSwitch |
| Product Type | Lysis Buffer L13 |
| Quantity | 800 mL |
| Shipping Condition | Room Temperature |
Frequently Asked Questions (FAQs)
We strongly recommend using the Elution Buffer provided in the kit, and do not recommend elution with water. If you need to elute in any other buffer, be sure to use a buffer of pH 8.5-9.0 for efficient DNA elution.
The filter kit is based upon a dual nested column design, which facilitates the fastest and simplest protocol for obtaining high quality plasmid (less than 10 EU/µg DNA). Modified wash buffer 1 removes more endotoxin.
The typical yield is 25 µg from a 5 mL culture for the ChargeSwitch Pro mini kit, 200 µg from a 25 mL culture for the midi kit, and 800 µg from a 100 mL culture from the maxi kit. The plasmid size range for the kits is typically 3-9 kb.
Based on a patented switchable chemistry, ChargeSwitch purification utilizes the charge on a solid surface for nucleic acid capture. At lower pH, the surface chemistry holds a positive charge and attracts nucleic acid. At higher pH, the surface chemistry is neutral and any bound nucleic acid is released.
For Research Use Only. Not for use in diagnostic procedures.