missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ Forensic DNA Purification Kit

Catalog No. p-4926399
Encompass
Change view
Click to view available options
No. of Reactions:
100 Preps
960 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
CS11200 100 Preps
CS1120010 960 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CS11200 Supplier Invitrogen™ Supplier No. CS11200
Only null left

Includes

ChargeSwitch Lysis Buffer (L13)
ChargeSwitch Magnetic Beads
Proteinase K (20 mg/mL in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl2, 50% glycerol)
ChargeSwitch Purification Buffer (N5)
ChargeSwitch Wash Buffer (W12)
ChargeSwitch Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

ChargeSwitch™ Forensic DNA Purification Kit uses magnetic bead technology to allow for rapid and efficient purification of genomic DNA from a variety of forensic samples, including poor or environmentally degraded samples, and is fully validated for Single Tandem Repeat (STR) analysis.

ChargeSwitch™ Forensic DNA Purification Kit uses magnetic bead technology to allow for rapid and efficient purification of genomic DNA from a variety of forensic samples, including poor or environmentally degraded samples, and is fully validated for Single Tandem Repeat (STR) analysis.

• Optimized for higher-purity DNA extraction
• Validated for STR profiling on diverse forensic samples-set
• Compatible with high-throughput protocols

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage

All components of the ChargeSwitch™ Forensic DNA Purification Kits are shipped at room temperature. Upon receipt, store the Proteinase K at 4°C. Store all other components at room temperature. All components are guaranteed stable for 6 months, if stored properly.

Components:
• ChargeSwitch™ Lysis Buffer (L13)
• ChargeSwitch™ Magnetic Beads
• Proteinase K (20 mg/mL in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl2, 50% glycerol)
• ChargeSwitch™ Purification Buffer (N5)
• ChargeSwitch™ Wash Buffer (W12)
• ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

Format Kit
Isolation Technology ChargeSwitch Chemistry
Sample Type Forensic Sample
For Use With (Application) Forensic
High-throughput Compatibility High-throughput Compatible
No. of Reactions 100 Preps
Product Line ChargeSwitch
Product Type Forensic DNA Purification Kit
Quantity 100 Preps
Shipping Condition Room Temperature
Target Genomic DNA
Test Time 15 min.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.