missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ Direct gDNA Purification Kit, 96-well

Catalog No. CS11205
Encompass
Change view
Click to view available options
Quantity:
96 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CS11205 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CS11205 Supplier Invitrogen™ Supplier No. CS11205

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

ChargeSwitch Plate Binding and lysis Buffer, ChargeSwitch Wash Buffers, ChargeSwitch Elution Buffer, ChargeSwitch Direct 96 gDNA Plates, and Plate Seals.

ChargeSwitch Direct 96 gDNA Kit is designed for rapid and efficient isolation of gDNA from fresh or frozen samples.

  • ChargeSwitch Direct 96 gDNA Kit is designed for rapid and efficient isolation of gDNA from fresh or frozen samples
  • Utilizes 96-well plates coated with ChargeSwitch nucleic acid-binding surface, whose ionic charge can be switched by changing pH of surrounding buffer
  • At low pH, positive charge of ChargeSwitch surface binds negatively charged nucleic acids
  • Proteins and other contaminants that do not bind are washed away, leaving pure DNA ready for downstream applications
  • Successful yields from very small samples volumes up to 10 μL blood or 2 x 104cells
  • Highly pure DNA isolated using gentle buffers—no guanidine or ethanol
  • No sample transfer that can lead to error or contamination—direct amplification in the same well as DNA binding without elution
  • Adaptable to your throughput needs—compatible with most automated Liquid handling platforms

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store at room temperature.
Format Kit
Isolation Technology ChargeSwitch Chemistry
Sample Type Blood, Cells
For Use With (Application) PCR, Southern Blotting, Sequencing, Cloning
High-throughput Compatibility High-throughput Compatible
Prep Scale < 100 μL Blood Samples (Small-scale)
Product Line ChargeSwitch
Product Type Direct gDNA Purification Kit
Quantity 96 Preps
Shipping Condition Room Temperature
Target Genomic DNA
Show More Show Less
I used water to elute my plasmid instead of the Elution Buffer with my ChargeSwitch kit, but saw very low plasmid yield. Why is this?

We strongly recommend using the Elution Buffer provided in the kit, and do not recommend elution with water. If you need to elute in any other buffer, be sure to use a buffer of pH 8.5-9.0 for efficient DNA elution.

What is the difference between the ChargeSwitch Pro kit and the Charge-Switch Pro filter kit?

The filter kit is based upon a dual nested column design, which facilitates the fastest and simplest protocol for obtaining high quality plasmid (less than 10 EU/µg DNA). Modified wash buffer 1 removes more endotoxin.

What is the typical yield when using the ChargeSwitch Pro Filter Plasmid Mini or Midi Kits?

The typical yield is 25 µg from a 5 mL culture for the ChargeSwitch Pro mini kit, 200 µg from a 25 mL culture for the midi kit, and 800 µg from a 100 mL culture from the maxi kit. The plasmid size range for the kits is typically 3-9 kb.

How does ChargeSwitch technology work?

Based on a patented switchable chemistry, ChargeSwitch purification utilizes the charge on a solid surface for nucleic acid capture. At lower pH, the surface chemistry holds a positive charge and attracts nucleic acid. At higher pH, the surface chemistry is neutral and any bound nucleic acid is released.

For Research Use Only. Not for use in diagnostic procedures.