missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BioPrime™ Array CGH Genomic Labeling System

Catalog No. 18095011
Encompass
Change view
Click to view available options
Quantity:
30 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18095011 30 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18095011 Supplier Invitrogen™ Supplier No. 18095011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

exo-Klenow fragment (40 U/μL), 2.5X random primer solution (octamers), control DNA (Salmon Sperm), stop buffer, and distilled water. The BioPrime™ Plus Systems and Modules contain the Alexa Fluor™ modified dNTP mixes or amine modified dNTP mixes and the Alexa Fluor™ NHS Esters. The standard BioPrime™ and BioPrime™ Plus Array CGH Genomic Labeling Systems also contain purification columns and purification buffers.

A high-performance labeling kit that enable reproducible labeling of genomic DNA samples for array-based Comparative Genomic Hybridization (CGH)

Using the BioPrime Plus Array CGH Genomic Labeling System, you can expect:
––High yields of fluorescently labeled genomic DNA
–signal-to-noise ratios
–detection of gene copy number variations.


These systems combine a highly concentrated exo-Klenow fragment of DNA polymerase I and random primers to effectively label genomic targets for sensitive genomic profiling experiments. Exo-Klenow is a mutant of the large fragment of the DNA polymerase I holoenzyme that has both 5' - 3' and 3' - 5' exonuclease activity removed. The lack of exonuclease activity makes this enzyme ideal for use in random priming protocols, enabling robust yields and efficient incorporation of Alexa Fluor™ AHA fluorescent nucleotides. In the BioPrime Plus Array CGH Genomic Labeling Systems, random octamers anneal to template DNA, providing priming sites for the exo- Klenow enzyme. Alexa Fluor AHA modified nucleotides (direct labeling) or amino-allyl modified nucleotides (indirect labeling) are incorporated as the polymerase extends from the priming sites. The labeled samples are then purified to remove contaminants prior to denaturing and hybridization to a microarray (direct labeling systems) or coupled to the Alexa Fluor NHS ester (indirect labeling systems) prior to hybridization. Available in a direct labeling format.

Array CGH, ChIP-on-Chip, Chromatin Biology, DNA and RNA Purification and Analysis, DNA Labeling, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Nucleic Acid Labeling and Oligo Synthesis, RNAi, Epigenetics and Non-Coding RNA Research

Order Info

Shipping Condition: Labeling Module, Dry Ice; Purification Module, Room Temperature

Specifications

Product Type Genomic Labeling System
Detection Method Fluorescence
Content And Storage Table 1 summarizes the components of the various BioPrime™ Array CGH Labeling Systems. The BioPrime™ Core Module is found in all standard BioPrime™ and BioPrime™ Plus Systems and Modules. It contains exo-Klenow fragment (40 U/ μl), 2.5X random primer solution (octamers), control DNA (Salmon Sperm), stop buffer, and distilled water. Store at -20°C. The BioPrime™ Plus Systems and Modules contain the Alexa Fluor™ modified dNTP mixes or amine modified dNTP mixes and the Alexa Fluor™ NHS Esters. Store at -20°C. The standard BioPrime™ and BioPrime™ Plus Array CGH Genomic Labeling Systems also contain purification columns and purification buffers. Store at room temperature. All components are guaranteed for 6 months when properly stored. See Table 1 for product component summary.
Final Product Type Probes (Labeled DNA)
Label or Dye Cy™3, Cy™5, Alexa Fluor™-Labeled Nucleotides
Includes Label or Dye No
Labeling Target Genomic DNA
Labeling Method Direct Labeling
Product Line BioPrime
For Use With (Application) Chromatin Biology
Quantity 30 Reactions
Show More Show Less
Can the PureLink columns provided to purify labeled cDNA probes be purchased separately? Which stand-alone columns are compatible?

The PureLink PCR purification system (Cat. Nos. K3100-01, K3100-02) can be used to purify labeled probe. Because tubes provided with stand-alone kit are not amber, be sure to protect your probe from light if you are doing direct labeling. Please contact Technical Support for the protocol.

Can the BioPrime Array CGH Genomic Labeling System be used for SNP analysis?

We do not recommend using the BioPrime Array CGH Genomic Labeling System to amplify genomic DNA for SNP analysis because the exo-Klenow polymerase has no proofreading activity.

For Research Use Only. Not for use in diagnostic procedures.