Learn More
MilliporeSigma™ Supelco™ Intermediate Polar Fused Silica Tubing
Ideal for use with injection solvents such as acetone, methylene chloride (dichloromethane), and toluene
Supplier:  MilliporeSigma™ Supelco™ 25339
 View more versions of this product
Catalog No. 111004673
Only null left
Description
Description
  • Use as transfer lines, guard columns, or retention gaps
  • Can be used to make custom columns
  • Ideal for use with injection solvents such as acetone, methylene chloride (dichloromethane), and toluene
  • Composition: Phenyl/Methyl
  • Deactivated
Specifications
Chemical Identifiers
Injection Solvents Such as Acetone, Methylene Chloride (Dichloromethane), and Toluene
Tubing
Use as Transfer Lines, Guard Columns, Retention Gaps, or Custom Columns
Silica
Specifications
Injection Solvents Such as Acetone, Methylene Chloride (Dichloromethane), and Toluene
Use as Transfer Lines, Guard Columns, Retention Gaps, or Custom Columns
5 m
Tubing
0.53 mm
Silica
Product Suggestions
Product Suggestions