missing translation for 'onlineSavingsMsg'
Learn More

Instrument Check Standard 4 For Environmental Analysis, SPEX CertiPrep™

Catalog No. CALMIX4100
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CALMIX4100 125 mL Triple-leached LDPE bottle
CALMIX4500 500 mL Triple-leached LDPE bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CALMIX4100 Supplier SPEX CertiPrep Group LLC Supplier No. CALMIX4100
Only null left

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
K50μg/mL
Al10μg/mL
As10μg/mL
Cu10μg/mL
Mn10μg/mL
Na10μg/mL
Ni10μg/mL
P10μg/mL
Pb10μg/mL
Sc10μg/mL
Zn10μg/mL
Ba1μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 2% HNO3
Quantity 125 mL
Grade Environmental
Concentration or Composition (by Analyte or Components) Components in 2% HNO3:
Al    •10Glc mug/mL
As    •10Glc mug/mL
Ba    •1Glc mug/mL
Cu    •10Glc mug/mL
Pb    •10Glc mug/mL
Mn    •10Glc mug/mL
Ni    •10Glc mug/mL
P    •10Glc mug/mL
K    •50Glc mug/mL
Sc    •10Glc mug/mL
Na    •10Glc mug/mL
Zn    •10Glc mug/mL
Chemical Name or Material Instrument Check Standard 4
Hazard Category Causes skin irritation, serious eye irritation
DOTInformation Hazardous
Recommended Storage Ambient

Product intended for laboratory use only