missing translation for 'onlineSavingsMsg'
Learn More

Instrument Check Standard 3, Ricca Chemical

Catalog No. RINSTCK3100
Catalog No. RINSTCK3100 Supplier Ricca Chemical Company Supplier No. RINSTCK3100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7664-39-3 , 7697-37-2 , 7732-18-5
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
PubChem CID 14917
ChEBI CHEBI:29228
IUPAC Name hydrogen fluoride
SMILES [H+].[F-]

Specifications

Packaging Natural LDPE Bottle
Odor Odorless
Chemical Name or Material Instrument Check Standard 3
ShelfLife 12 months