missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Instrument Calibration Standard 1 For ICP/MS, SPEX CertiPrep™
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ag | 20μg/mL |
| Al | 20μg/mL |
| As | 20μg/mL |
| Ba | 20μg/mL |
| Be | 20μg/mL |
| Cd | 20μg/mL |
| Co | 20μg/mL |
| Cr | 20μg/mL |
| Cu | 20μg/mL |
| Mn | 20μg/mL |
| Mo | 20μg/mL |
| Ni | 20μg/mL |
| Pb | 20μg/mL |
| Sb | 20μg/mL |
| Se | 20μg/mL |
| Th | 20μg/mL |
| Tl | 20μg/mL |
| U | 20μg/mL |
| V | 20μg/mL |
| Zn | 20μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Packaging | Triple-leached LDPE bottle |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| Technique | ICP-MS |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Solution Type | Standard Solution |
| Type | ICP/ICP-MS Standard |
| Solvent or Matrix | 5% HNO3/tr Tart. Acid |
| Quantity | 125 mL |
| Grade | ICP/ICP-MS |
| Concentration or Composition (by Analyte or Components) | Each component at 20μg/mL in 2% HNO3/tr. Tartaric Acid:, Aluminum, Antimony, Arsenic, Barium, Beryllium, Cadmium, Chromium, Cobalt, Copper, Lead, Manganese, Molybdenum, Nickel, Selenium, Silver, Thallium, Thorium, Uranium, Vanadium, Zinc |
| Show More |
Safety and Handling
| Hazard Category | Danger: Causes severe skin burns and eye damage Do not breathe dusts or mists. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| DOTInformation | Hazardous |
| Recommended Storage | Ambient |
Product intended for laboratory use only