missing translation for 'onlineSavingsMsg'
Learn More

Instrument Calibration Standard 1 For CLP, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC ICAL1

Catalog No. ICAL-1


Only null left

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ca5000μg/mL
K5000μg/mL
Mg5000μg/mL
Na5000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Instrument Calibration Standard 1
7697-37-2
HNO3
GC-MS
Certified Reference Material
O[N+]([O-])=O
nitric acid
125 mL
CHEBI:48107
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00011349
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
GRYLNZFGIOXLOG-UHFFFAOYSA-N
5% HNO3
63.01
944
Each component at 5000μg/mL in 5% HNO3:, Calcium, Magnesium, Potassium, Sodium

Chemical Identifiers

7697-37-2
63.01
GRYLNZFGIOXLOG-UHFFFAOYSA-N
CHEBI:48107
O[N+]([O-])=O
HNO3
MFCD00011349
944
nitric acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes severe skin burns and eye damage

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS Documents

Documents

Promotions

Promotions

Product intended for laboratory use only