missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Inserts for 1.5 mL Large Opening Vials

Allow the use of smaller sample volumes with standard autosampler vials.

$201.00

Specifications

Volume (Metric) 0.1 mL
View More Specs

Products 1
Catalog Number Mfr. No. Insert Type Price Quantity & Availability  
Catalog Number Mfr. No. Insert Type Price Quantity & Availability  
111010584
View Documents Promotion Details
MilliporeSigma™ Supelco™
SU860066
Conical with Bottom Spring
Each for $201.00
Only null left
 
Description

Description

  • Vial inserts for 1.5 mL large opening vials [0.1 mL, clear glass conical (with bottom spring)] allow the use of smaller sample volumes with standard autosampler vials.
  • Inserts are generally chosen based on the inner diameter of the vial neck.
  • Large opening vials (6 mm ID) require a 6 mm OD insert.
  • Clear glass conical inserts used with CTC LC PAL autosamplers.
Specifications

Specifications

0.1 mL
Videos
SDS
Documents

Documents