missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Inosine, 98+%

Catalog No. AAA1445922
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1445922 100 g
AAA1445906 5 g
AAA1445914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1445922 Supplier Thermo Scientific Chemicals Supplier No. A1445922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58-63-9 | C10H12N4O5 | 268.23 g/mol

Suppresses the increase of glucose and insulin in the blood

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58-63-9
Molecular Formula C10H12N4O5
Molecular Weight (g/mol) 268.23
MDL Number MFCD00066770
InChI Key UGQMRVRMYYASKQ-YPLCUDRINA-N
Synonym inosine, hypoxanthosine, ribonosine, atorel, oxiamin, trophicardyl, selfer, pantholic-l, panholic-l, hypoxanthine riboside
PubChem CID 6021
ChEBI CHEBI:17596
IUPAC Name 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1N=CNC2=O

Specifications

Melting Point 212°C to 215°C (decomposition)
Quantity 100 g
Beilstein 624889
Merck Index 14,4975
Optical Rotation −52° (c=1 in Water)
Formula Weight 268.23
Percent Purity ≥98%
Chemical Name or Material Inosine
EINECSNumber 200-390-4
RTECSNumber NW7460000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only