accessibility menu, dialog, popup

UserName

N(R)N(R)ON(S)(R)HNOOHHOHO

CAS RN 58-63-9

IUPAC Name: 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Synonyms: 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-purin-6-one
Molecular Weight (g/mol): 268.23
Molecular Formula: C10H12N4O5
InChi Key: UGQMRVRMYYASKQ-YPLCUDRINA-N
SMILES: OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1N=CNC2=O
Percent Purity 
  • (3)
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (7)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (6)